Merck
Incorporation of ophiobolin a into novel chemoembolization particles for cancer cell treatment.
Pharmaceutical research (2014-05-06)
Rachel Morrison, Chris Gardiner, Antonio Evidente, Robert Kiss, Helen Townley
ZUSAMMENFASSUNG
To design and synthesize chemoembolization particles for the delivery of Ophiobolin A (OphA), a promising fungal-derived chemotherapeutic, directly at the tumour location. To investigate cell death mechanism of OphA on a Rhabdomyosarcoma cancer (RD) cell line. Rhabdomyosarcoma is the most common soft tissue sarcoma in children; with a 5-year survival rate of between 30 and 65%. Multimodal chemoembolization particles were prepared by sintering mesoporous silica nanoparticles, prepared by the sol-gel method, onto the surface of polystyrene microspheres, prepared by suspension copolymerisation. The chemoembolization particles were subsequently loaded with OphA. The effects of OphA in vitro were characterised by flow cytometry and nanoparticle tracking analysis (NanoSight). High loading of OphA onto the chemoembolization particles was achieved. The subsequent release of OphA onto RD cells in culture showed a 70% reduction in cell viability. OphA caused RD cells to round up and their membrane to bleb and caused cell death via apoptosis. OphA caused both an increase in the number of microvesicles produced and an increase in DNA content within these microvesicles. The prepared chemoembolization particles showed good efficacy against RD cells in culture.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Tetraethylorthosilikat, reagent grade, 98%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
2,2′-Azobis(2-Methylpropionitril), 98%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, Molecular Biology, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Tetraethylorthosilikat, ≥99.0% (GC)
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Styrol, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
SAFC
L-Glutamin
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, BioXtra, ≥99%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC