Merck
  • Ultrahigh-performance liquid chromatography/tandem mass spectrometry method for evaluating enzyme activity and screening inhibitors of cyclooxygenase-2.
Ultrahigh-performance liquid chromatography/tandem mass spectrometry method for evaluating enzyme activity and screening inhibitors of cyclooxygenase-2.
Rapid communications in mass spectrometry : RCM (2015-01-07)
Hongbin Zhu, Xiaoyu Zhuang, Shu Liu, Zifeng Pi, Fengrui Song, Na Lin, Zhiqiang Liu
ZUSAMMENFASSUNG
Prostaglandin E2 is an important biomarker in many biological systems. The development of sensitive and reliable analytical methods for monitoring PGE2 contents in various samples is of great interest. Here we developed an improved method for evaluating the enzyme activity and screening COX-2 inhibitors using ultrahigh-performance liquid chromatography/tandem mass spectrometry (UPLC/MS/MS) combined with PGE2 derivatization. Girard's reagent-T was used as the derivatization reagent and the reaction conditions were optimized. The established method was performed to screen the COX-2 inhibitors from effective constituents of herbs and detect the concentration of PGE2 in biological tissue samples (liver and kidney). The IC50 values of celecoxib, rofecoxib, sinomenine, bulleyaconitine A, tetrandrine, fangchinoline, berberine hydrochloride and sophocarpidine towards COX-2 were determined. This method improves the quantitative ability for PGE2 , including the linearly dependent coefficient, linearity range and limit of detection. After derivatization, the derivatized PGE2 could be detected in positive ion mode of electrospray ionization (ESI), which improves the detection sensitivity 10-fold compared to that of the direct detection of underivatized PGE2 in negative ESI mode. Besides the positive control, sinomenine (IC50 =113 μM) and bulleyaconitine A (IC50=53 μM) were found to be potent COX-2 inhibitors. All the results indicate that the present derivatization quantification method of PGE2 could be used as the detection method of COX-2 enzyme activity and as the screening method for COX-2 inhibitors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumbicarbonat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
(−)-Epinephrin
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammoniumbicarbonat, BioUltra, ≥99.5% (T)
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Arachidonsäure, >95.0% (GC)
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%