Merck
  • Identification of protein O-glycosylation site and corresponding glycans using liquid chromatography-tandem mass spectrometry via mapping accurate mass and retention time shift.
Identification of protein O-glycosylation site and corresponding glycans using liquid chromatography-tandem mass spectrometry via mapping accurate mass and retention time shift.
Journal of chromatography. A (2014-12-03)
Li-Juan Huang, Jen-Hui Lin, Jung-Heng Tsai, Yen-Yin Chu, Yen-Wen Chen, Shun-Li Chen, Shu-Hui Chen
ZUSAMMENFASSUNG
We reported an improved combinatorial approach for identifying site-specific O-glycosylation using both glycan cleaved and non-cleaved methods. In this approach, a non-reducing β-elimination kit coupled with non-specific enzymes performed efficient digestion, O-glycan cleavage, and partial dephosphorylation without significant side reactions, thus enabling an automatic database search for the cleaved O-glycosylation or serine/threonine (S/T) phosphorylation sites. From the same sample concurrently prepared without β-elimination, the corresponding intact O-glycopeptides were mapped by accurate precursor ion mass using an in-house glycan database majorly composed of GalNAc (mucin-type) core and the retention-time shift (ΔRt). Each glycopeptide assignment was verified by the detection of glycan-specific fragments using collision-induced dissociation (CID) to estimate False Discovery Rate (FDR). Using fetuin as a model, all identified S/T elimination sites were matched to multiple intact glycopeptides with a 31% FDR. This considerably reduced to 0% FDR by ΔRt filtering. This approach was then applied to a protein mixture composed of therapeutic Factor IX and Enbrel(®) mixed with fetuin and kappa-casein. A total of 26 glycosylation sites each of which corresponds to 1-4 glycans were positively mapped and confirmed. The FDR decreased from 33% to 3.3% by ΔRt filtering and exclusion of repeated peptide tags that covered the same glycosylation sites. Moreover, the phosphorylation and O-glycosylation on the same site such as T159 of Factor IX and T170 of kappa-casein were able to be unambiguously differentiated. Thus, our approach is useful for in-depth characterization of site-specific O-glycosylation of a simple mixture such as protein-based therapeutics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitril, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumbicarbonat, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Guanidin -hydrochlorid, Molecular Biology, ≥99%
Sigma-Aldrich
Jodacetamid, BioUltra
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Jodacetamid, Single use vial of 56 mg
Sigma-Aldrich
Acetonitril, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Jodacetamid, ≥99% (NMR), crystalline
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Guanidin -hydrochlorid, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Guanidin -hydrochlorid, ≥98%
Sigma-Aldrich
Ammoniumbicarbonat, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Guanidin -hydrochlorid -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Guanidin -hydrochlorid -Lösung, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Guanidin -hydrochlorid, ≥99.0% (AT)
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Supelco
Acetonitril (unverdünnt), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Guanidin -hydrochlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
SAFC
Jodacetamid
Supelco
Acetonitril, analytical standard
Sigma-Aldrich
Guanidin -hydrochlorid, anhydrous, free-flowing, Redi-Dri, ≥99%