Merck
  • Optimization of a chromatographic stationary phase based on gellan gum using central composite design.
Optimization of a chromatographic stationary phase based on gellan gum using central composite design.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-03-25)
A I C Gonçalves, L A Rocha, J M L Dias, L A Passarinha, A Sousa
ZUSAMMENFASSUNG
To develop a new stationary phase of easy production, low cost, biocompatible, biodegradable and low unspecific adsorption, a three-dimensional network was prepared by combining the natural polysaccharide of gellan with divalent cations. The stability of this cation exchange chromatographic matrix was optimized by using an experimental design tool. The optimal conditions proposed for the gellan gel formulation were 48mM ZnSO4, 0% DMF, 25°C, 0.75% gellan and 0.5h. The applicability of gellan matrix was tested by chromatographic assays with three model proteins (bovine serum albumin (BSA), α-chymotripsin and lysozyme). The results showed that the retention occurred in function of the net charge of each protein in MES buffer pH 6.2 and the elution was performed by increase of ionic strength to 750mM NaCl in MES buffer pH 6.2. Lysozyme was the more retained protein due to its positive charge more effective than α-chymotripsin, while BSA did not interact with the matrix due to its negative charge at these conditions. Dynamic binding capacity assays were accomplished to characterize this matrix and to compare with commercial resins. The values of dynamic binding capacity from gellan gel were 3.9mg/mL and 17.4mg/mL, at 10% and 50% of breakthrough, respectively. In this way, gellan gel might be a promising chromatographic matrix to explore ionic interactions and to be applied in different purification strategies, getting the best benefit from its use at low cost.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, suitable for electrophoresis, ≥99.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ≥99.5%, purified by redistillation
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylendiamin, ReagentPlus®, 99%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Acrylamid -Lösung, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Dimethylformamid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%