Merck
  • An aqueous extract of Limoniastrum guyonianum gall induces anti-tumor effects in melanoma-injected mice via modulation of the immune response.
An aqueous extract of Limoniastrum guyonianum gall induces anti-tumor effects in melanoma-injected mice via modulation of the immune response.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-04-08)
Mounira Krifa, Ines Skandrani, Antonio Pizzi, Nouha Nasr, Zied Ghedira, Nadia Mustapha, Kamel Ghedira, Leila Chekir-Ghedira
ZUSAMMENFASSUNG
The objectives of this study were to evaluate the in vitro and in vivo anti-tumor potential of the aqueous gall extract (G extract) from Limoniastrum guyonianum and to elucidate its immunological mechanisms, in part, by assessing its effects on the growth of transplanted tumors and the immune response in these tumor-bearing mice. Here, mice were inoculated with B16F10 mouse tumor cells and then treated intraperitoneally with G extract at 25 or 50 mg extract/kg BW for 7, 14, or 21 days. At each timepoint, effects of the extract on the tumor growth, splenocytes proliferation, NK cell activity, and CTL activity among splenocytes isolated from the mice were measured. G extract-induced tumor growth inhibition was associated with characteristic apoptotic changes in the tumor cells, like nuclear condensation. In addition, the extract inhibited melanin synthesis and tyrosinase activity among melanoma cells in a concentration-related manner. G extract did not only significantly inhibit the growth of the transplantable tumor, but also remarkably increased splenocytes proliferation and both NK and CTL activities in tumor-bearing mice. The extract was also seen to have promoted lysosomal activity of host macrophages and gave rise to enhanced cellular anti-oxidant activity in several cell types in mice.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
3,4-Dihydroxy-L-Phenylalanin, ≥98% (TLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Tanninsäure, ACS reagent
SAFC
L-Glutamin
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tanninsäure
USP
Levodopa, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Supelco
Dimethylsulfoxid, analytical standard
Supelco
Dimethylsulfoxid, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethylsulfoxid, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Glutamin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Tanninsäure, Source: Chinese natural gall nuts
Levodopa, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland