Merck
  • Localization of a prostate cancer predisposition gene to an 880-kb region on chromosome 22q12.3 in Utah high-risk pedigrees.
Localization of a prostate cancer predisposition gene to an 880-kb region on chromosome 22q12.3 in Utah high-risk pedigrees.
Cancer research (2006-10-19)
Nicola J Camp, James M Farnham, Lisa A Cannon-Albright
ZUSAMMENFASSUNG
Chromosome 22q has become recently a region of interest for prostate cancer. We identified previously a logarithm of odds (LOD) of 2.42 at chromosome 22q12.3. Additionally, this region has been noted by eight other studies, with linkage evidence ranging from LOD of 1.50 to 3.57. Here, we do fine mapping and localization of the region using a pedigree-specific recombinant mapping approach in 14 informative, high-risk Utah pedigrees. These 14 pedigrees were chosen because they were either "linked" or "haplotype-sharing" pedigrees or both. "Linked" pedigrees were those with significant pedigree-specific linkage evidence (LOD, >0.588; P < 0.05) to the 22q12.3 region, regardless of the number of prostate cancer cases sharing the segregating haplotype. "Haplotype-sharing" pedigrees were those with at least five prostate cancer cases sharing a segregating haplotype in the 22q12.3 region, regardless of the linkage evidence. In each pedigree, the most likely haplotype configuration (in addition to the multipoint LOD graph for linked pedigrees) was used to infer the position of recombinant events and delimit the segregating chromosomal segment in each pedigree. These pedigree-specific chromosomal segments were then overlaid to form a consensus recombinant map across all 14 pedigrees. Using this method, we identified a 881,538-bp interval at 22q12.3, between D22S1265 and D22S277, which is the most likely region that contains the 22q prostate cancer predisposition gene. The unique Utah extended high-risk pedigree resource allows this powerful localization approach in pedigrees with evidence for segregating predisposition to prostate cancer. We are mutation screening candidate genes in this region to identify specific genetic variants segregating in these pedigrees.