Merck
  • Listeria monocytogenes-carrying consortia in food industry. Composition, subtyping and numerical characterisation of mono-species biofilm dynamics on stainless steel.
Listeria monocytogenes-carrying consortia in food industry. Composition, subtyping and numerical characterisation of mono-species biofilm dynamics on stainless steel.
International journal of food microbiology (2015-05-24)
Pedro Rodríguez-López, Paula Saá-Ibusquiza, Maruxa Mosquera-Fernández, Marta López-Cabo
ZUSAMMENFASSUNG
In order to find out how real Listeria monocytogenes-carrying biofilms are in industrial settings, a total of 270 environmental samples belonging to work surfaces from fish (n = 123), meat (n = 75) and dairy industries (n = 72) were analysed in order to detect L. monocytogenes. 12 samples were positive for L. monocytogenes and a total of 18 different species were identified as accompanying microbiota in fish and meat industry. No L. monocytogenes was found in samples from dairy industry. Molecular characterisation combining results of AscI and ApaI macrorestriction PFGE assays yielded 7 different subtypes of L. monocytogenes sharing in 71.43% of cases the same serogroup (1/2a-3a). Results from dynamic numerical characterisation between L. monocytogenes monospecies biofilms on stainless steel (SS) using MATLAB-based tool BIOFILMDIVER demonstrated that except in isolate A1, in which a significant increase in the percentage of covered area (CA), average diffusion distance (ADD) and maximum diffusion distance (MDD) was observed after 120 h of culture, no significant differences were observed in the dynamics of the rest of the L. monocytogenes isolates. Quantitative dual-species biofilm association experiments performed on SS indicated that L. monocytogenes cell counts presented lower values in mixed-species cultures with certain species at 24 and 48 h compared with mono-species culture. However, they remained unaltered after 72 h except when co-cultured with Serratia fonticola which presented differences in all sampling times and was also the dominant species within the dual-species biofilm. When considering frequency of appearance of accompanying species, an ecological distribution was demonstrated as Escherichia coli appeared to be the most abundant in fish industry and Carnobacterium spp. in meat industry.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Ethidiumbromid, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Ethidiumbromid, ~95% (HPLC)
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ethidiumbromid -Lösung, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O