Merck
  • Synthesis and evaluation of novel 1H-pyrrolo[2,3-b]pyridine-5-carboxamide derivatives as potent and orally efficacious immunomodulators targeting JAK3.
Synthesis and evaluation of novel 1H-pyrrolo[2,3-b]pyridine-5-carboxamide derivatives as potent and orally efficacious immunomodulators targeting JAK3.
Bioorganic & medicinal chemistry (2015-06-14)
Yutaka Nakajima, Takayuki Inoue, Kazuo Nakai, Koichiro Mukoyoshi, Hisao Hamaguchi, Keiko Hatanaka, Hiroshi Sasaki, Akira Tanaka, Fumie Takahashi, Shigeki Kunikawa, Hiroyuki Usuda, Ayako Moritomo, Yasuyuki Higashi, Masamichi Inami, Shohei Shirakami
ZUSAMMENFASSUNG
Janus kinases (JAKs) regulate various inflammatory and immune responses and are targets for the treatment of inflammatory and immune diseases. As a novel class of immunomodulators targeting JAK3, 1H-pyrrolo[2,3-b]pyridine-5-carboxamide derivatives are promising candidates for treating such diseases. In chemical modification of lead compound 2, the substitution of a cycloalkyl ring for an N-cyanopyridylpiperidine in C4-position was effective for increasing JAK3 inhibitory activity. In addition, modulation of physical properties such as molecular lipophilicity and basicity was important for reducing human ether-a-go-go-related gene (hERG) inhibitory activity. Our optimization study gave compound 31, which exhibited potent JAK3 inhibitory activity as well as weak hERG inhibitory activity. In cellular assay, 31 exhibited potent immunomodulating effect on IL-2-stimulated T cell proliferation. In a pharmacokinetic study, good metabolic stability and oral bioavailability of 31 were achieved in rats, dogs, and monkeys. Further, 31 prolonged graft survival in an in vivo rat heterotopic cardiac transplant model.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Dodecan, anhydrous, ≥99%
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O