Merck
  • Image quality at low tube voltage (70 kV) and sinogram-affirmed iterative reconstruction for computed tomography in infants with congenital heart disease.
Image quality at low tube voltage (70 kV) and sinogram-affirmed iterative reconstruction for computed tomography in infants with congenital heart disease.
Pediatric radiology (2015-06-28)
Motoo Nakagawa, Yoshiyuki Ozawa, Keita Sakurai, Masashi Shimohira, Kazuya Ohashi, Miki Asano, Sachiko Yamaguchi, Yuta Shibamoto
ZUSAMMENFASSUNG
Lower tube voltage has advantages for CT angiography, such as improved contrast To evaluate the image quality of low-voltage (70 kV) CT for congenital heart disease and the ability of sinogram-affirmed iterative reconstruction to improve image quality. Forty-six children with congenital heart disease (median age: 109 days) were examined using dual-source CT. Scans were performed at 80 kV and 70 kV in 21 and 25 children, respectively. A nonionic iodinated contrast medium (300 mg I/ml) was used for the 80-kV protocol. The contrast medium was diluted to 75% (225 mgI/mL) with saline for the 70-kV protocol. Image noise was measured in the two protocols for each group by extracting the standard deviations of a region of interest placed on the descending aorta. We then determined whether sinogram-affirmed iterative reconstruction reduced the image noise at 70 kV. There was more noise at 70 kV than at 80 kV (29 ± 12 vs 20 ± 4.8; P < 0.01). Sinogram-affirmed iterative reconstruction with grade 4 strength settings improved the noise (20 ± 5.9; P < 0.01) for the 70-kV group. Sinogram-affirmed iterative reconstruction improved the image quality of CT in congenital heart disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Histodenz, nonionic density gradient medium
Preise und Verfügbarkeit sind derzeit nicht verfügbar.