Merck
Use of DMPC and DSPC lipids for verapamil and naproxen permeability studies by PAMPA.
Drug development and industrial pharmacy (2014-02-27)
M J Alvarez-Figueroa, B C Contreras-Garrido, M A Soto-Arriaza
ZUSAMMENFASSUNG
Verapamil and naproxen Parallel Artificial Membrane Permeability Assay (PAMPA) permeability was studied using lipids not yet reported for this model in order to facilitate the quantification of drug permeability. These lipids are 1,2-distearoyl-sn-glycero-3-phosphatidylcholine (DSPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) and an equimolar mixture of DMPC/DSPC, both in the absence and in the presence of 33.3 mol% of cholesterol. PAMPA drug permeability using the lipids mentioned above was compared with lecithin-PC. The results show that verapamil permeability depends on the kind of lipid used, in the order DMPC > DMPC/DSPC > DSPC. The permeability of the drugs was between 1.3 and 3.5-times larger than those obtained in lecithin-PC for all the concentrations of the drug used. Naproxen shows similar permeability than verapamil; however, the permeability increased with respect to lecithin-PC only when DMPC and DMPC/DSPC were used. This behavior could be explained by a difference between the drug net charge at pH 7.4. On the other hand, in the presence of cholesterol, verapamil permeability increases in all lipid systems; however, the relative verapamil permeability respect to lecithin-PC did not show any significant increase. This result is likely due to the promoting effect of cholesterol, which is not able to compensate for the large increase in verapamil permeability observed in lecithin-PC. With respect to naproxen, its permeability value and relative permeability respect lecithin-PC not always increased in the presence of cholesterol. This result is probably attributed to the negative charge of naproxen rather than its molecular weight. The lipid systems studied have an advantage in drug permeability quantification, which is mainly related to the charge of the molecule and not to its molecular weight or to cholesterol used as an absorption promoter.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Dodecan, anhydrous, ≥99%
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthalinessigsäure, 98%
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®