Merck
  • CCR8 regulates contact hypersensitivity by restricting cutaneous dendritic cell migration to the draining lymph nodes.
CCR8 regulates contact hypersensitivity by restricting cutaneous dendritic cell migration to the draining lymph nodes.
International immunology (2014-10-27)
Rikio Yabe, Kenji Shimizu, Soichiro Shimizu, Satoe Azechi, Byung-Il Choi, Katsuko Sudo, Sachiko Kubo, Susumu Nakae, Harumichi Ishigame, Shigeru Kakuta, Yoichiro Iwakura
ZUSAMMENFASSUNG
Allergic contact dermatitis (ACD) is a typical occupational disease in industrialized countries. Although various cytokines and chemokines are suggested to be involved in the pathogenesis of ACD, the roles of these molecules remain to be elucidated. CC chemokine receptor 8 (CCR8) is one such molecule, of which expression is up-regulated in inflammatory sites of ACD patients. In this study, we found that Ccr8(-/-) mice developed severer contact hypersensitivity (CHS) responses to 2,4-dinitrofluorobenzene, a murine model of ACD, compared with wild-type mice. T cells from Ccr8(-/-) mice showed enhanced proliferative recall responses and Th1 and Th17 cell populations were expanded in these mice. However, CHS responses were similar between SCID mice adoptively transferred with Ccr8(-/-) and wild-type T cells, suggesting that CCR8 in T cells is not responsible for the exacerbation of CHS. Notably, skin-resident dendritic cells (DCs), such as Langerhans cells and dermal DCs, and inflammatory DCs were highly accumulated in lymph nodes (LNs) of Ccr8(-/-) mice after sensitization. Consistent with this, Ccr8(-/-) antigen-presenting cells readily migrated from the skin to the draining LNs after sensitization. These observations suggest that CCR8 negatively regulates migration of cutaneous DCs from the skin to the draining LNs in CHS by keeping these cells in the skin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
1-Fluor-2,4-Dinitrobenzol, ≥99%
Sigma-Aldrich
1-Fluor-2,4-Dinitrobenzol, purum p.a., ≥98.0% (GC)
Sigma-Aldrich
Azodicarbonsäure-diethylester -Lösung, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
N-Acetyl-D-lactosamin, ≥98% (TLC)
Sigma-Aldrich
Thrombopoietin human, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, ≥98% (SDS-PAGE and HPLC)
Sigma-Aldrich
Thrombopoietin from mouse, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, >97% (SDS-PAGE)