Merck
  • Pregnancy Rates to Fixed Embryo Transfer of Vitrified IVP Bos indicus, Bos taurus or Bos indicus × Bos taurus Embryos.
Pregnancy Rates to Fixed Embryo Transfer of Vitrified IVP Bos indicus, Bos taurus or Bos indicus × Bos taurus Embryos.
Reproduction in domestic animals = Zuchthygiene (2015-08-19)
L S R Marinho, B V Sanches, C O Rosa, J H Tannura, A G Rigo, A C Basso, J H F Pontes, M M Seneda
ZUSAMMENFASSUNG
The pregnancy rates obtained after the transfer of cryopreserved in vitro-produced (IVP) embryos are usually low and/or inconsistent. The objective of this study was to evaluate the pregnancy rates of Holstein, Gyr and Holstein × Gyr cattle after the transfer of vitrified IVP embryos produced with X-sorted sperm. Seventy-two Gyr and 703 Holstein females were subjected to ovum pickup (OPU) sessions, followed by in vitro embryo production using semen from sires of the same breeds. Embryos (1636 Holstein, 241 Gyr and 1515 Holstein × Gyr) were exposed to forskolin for 48 h prior to vitrification. The pregnancy rate achieved with Gyr dam and sire was 46.1%, which was similar (p = 0.11) to that of Holstein dam and Gyr sire (40.3%). Crossing Gyr dams with Holstein sires resulted in a pregnancy rate of 38.9% and did not differ (p = 0.58) from the pregnancy rate obtained with the cross between Holstein dams and Gyr sires. The rate obtained with Holstein dam and sire was 32.5%. The average pregnancy rate was 36.6%, and no difference was found in the proportion of female foetuses (88.8%, in average) among breeds (p > 0.05). In conclusion, transfer of cryopreserved X-sorted embryos represents an interesting choice for dairy cattle. Despite the small differences between pregnancy rates, we highlight the efficiency of this strategy for all of the racial groups studied.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Ethylenglycol, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Heparin Natriumsalz aus Schweinedarm, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenglycol, anhydrous, 99.8%
Sigma-Aldrich
Ethylenglycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Progesteron, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Progesteron, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
β-Estradiol, meets USP testing specifications
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Progesteron, meets USP testing specifications
Sigma-Aldrich
Ethylenglykol 5 M -Lösung
Sigma-Aldrich
Progesteron, γ-irradiated, BioXtra, suitable for cell culture