Merck
  • Influence of temperature on thyroid hormone signaling and endocrine disruptor action in Rana (Lithobates) catesbeiana tadpoles.
Influence of temperature on thyroid hormone signaling and endocrine disruptor action in Rana (Lithobates) catesbeiana tadpoles.
General and comparative endocrinology (2015-01-15)
S Austin Hammond, Nik Veldhoen, Caren C Helbing
ZUSAMMENFASSUNG
Thyroid hormones (THs) are essential for normal growth, development, and metabolic control in vertebrates. Their absolute requirement during amphibian metamorphosis provides a powerful means to detect and assess the impact of environmental contaminants on TH signaling in the field and laboratory. As poikilotherms, frogs can experience considerable temperature fluctuations. Previous work demonstrated that low temperature prevents precocious TH-dependent induction of metamorphosis. However, a shift to a permissive higher temperature allows resumption of the induced metamorphic program regardless of whether or not TH remains. We investigated the impact of temperature on the TH-induced gene expression programs of premetamorphic Rana (Lithobates) catesbeiana tadpoles following a single injection of 10pmol/g body wet weight 3,3',5-triiodothyronine (T3). Abundance profiles of several T3-responsive mRNAs in liver, brain, lung, back skin, and tail fin were characterized under permissive (24°C), nonpermissive (5°C), or temperature shift (5-24°C) conditions. While responsiveness to T3 was retained to varying degrees at nonpermissive temperature, T3 modulation of thibz occurred in all tissues at 5°C suggesting an important role for this transcription factor in initiation of T3-dependent gene expression programs. Low temperature immersion of tadpoles in water containing 10nM T3 and the nonsteroidal anti-inflammatory drug, ibuprofen, or the antimicrobial agent, triclosan, perturbed some aspects of the gene expression programs of tail fin and back skin that was only evident upon temperature shift. Such temporal uncoupling of chemical exposure and resultant biological effects in developing frogs necessitates a careful evaluation of environmental temperature influence in environmental monitoring programs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl-3-aminobenzoat -methansulfonat, 98%
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Sigma-Aldrich
Ascarit®, Sodium hydroxide-coated silica, 8-20 mesh
Sigma-Aldrich
Ascarit®, Sodium hydroxide-coated silica, 20-30 mesh
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C