Merck
Oxidative Stress and Nano-Toxicity Induced by TiO2 and ZnO on WAG Cell Line.
PloS one (2015-05-27)
Akhilesh Dubey, Mukunda Goswami, Kamalendra Yadav, Dharmendra Chaudhary
ZUSAMMENFASSUNG
Metallic nanoparticles are widely used in cosmetics, food products and textile industry. These particles are known to cause respiratory toxicity and epithelial inflammation. They are eventually released to aquatic environment necessitating toxicity studies in cells from respiratory organs of aquatic organisms. Hence, we have developed and characterized a new cell line, WAG, from gill tissue of Wallago attu for toxicity assessment of TiO2 and ZnO nanoparticles. The efficacy of the cell line as an in vitro system for nanoparticles toxicity studies was established using electron microscopy, cytotoxicity assays, genotoxicity assays and oxidative stress biomarkers. Results obtained with MTT assay, neutral red uptake assay and lactate dehydrogenase assay showed acute toxicity to WAG cells with IC50 values of 25.29 ± 0.12, 34.99 ± 0.09 and 35.06 ± 0.09 mg/l for TiO2 and 5.716 ± 0.1, 3.160 ± 0.1 and 5.57 ± 0.12 mg/l for ZnO treatment respectively. The physicochemical properties and size distribution of nanoparticles were characterized using electron microscopy with integrated energy dispersive X-ray spectroscopy and Zetasizer. Dose dependent increase in DNA damage, lipid peroxidation and protein carbonylation along with a significant decrease in activity of Superoxide Dismutase, Catalase, total Glutathione levels and total antioxidant capacity with increasing concentration of exposed nanoparticles indicated that the cells were under oxidative stress. The study established WAG cell line as an in vitro system to study toxicity mechanisms of nanoparticles on aquatic organisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Ethyl-3-aminobenzoat -methansulfonat, 98%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis