Merck
  • Synthesis of [(18) F] 4-amino-N-(3-chloro-4-fluorophenyl)-N'-hydroxy-1,2,5-oxadiazole-3-carboximidamide (IDO5L): a novel potential PET probe for imaging of IDO1 expression.
Synthesis of [(18) F] 4-amino-N-(3-chloro-4-fluorophenyl)-N'-hydroxy-1,2,5-oxadiazole-3-carboximidamide (IDO5L): a novel potential PET probe for imaging of IDO1 expression.
Journal of labelled compounds & radiopharmaceuticals (2015-02-19)
Xuan Huang, Robert J Gillies, Haibin Tian
ZUSAMMENFASSUNG
To synthesize (18) F-labeled positron emission tomography (PET) ligands, reliable labeling techniques inserting (18) F into a target molecule are necessary. The (18) F-fluorobenzene moiety has been widely utilized in the synthesis of (18) F-labeled compounds. The present study utilized [(18) F]-labeled aniline as intermediate in [(18) F]-radiolabeling chemistry for the facile radiosynthesis of 4-amino-N-(3-chloro-4-fluorophenyl)-N'-hydroxy-1,2,5-oxadiazole-3-carboximidamide ([(18) F]IDO5L) as indoleamine 2,3-dioxygenase 1 (IDO1) targeted tracer. IDO5L is a highly potent inhibitor of IDO1 with low nanomolar IC50 . [(18) F]IDO5L was synthesized via coupling [(18) F]3-chloro-4-fluoroaniline with carboximidamidoyl chloride as a potential PET probe for imaging IDO1 expression. Under the optimized labeling conditions, chemically and radiochemically pure (>98%) [(18) F]IDO5L was obtained with specific radioactivity ranging from 11 to 15 GBq/µmol at the end of synthesis within ~90 min, and the decay-corrected radiochemical yield was 18.2 ± 2.1% (n = 4).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Kaliumcarbonat, 99.995% trace metals basis
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Natriumiodid, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Kaliumcarbonat, powder, 99.99% trace metals basis
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Wasser, endotoxin, free
Sigma-Aldrich
Natriumiodid, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Kaliumcarbonat, meets USP testing specifications
Sigma-Aldrich
Natriumiodid, ≥99.99% trace metals basis
Sigma-Aldrich
Kaliumcarbonat, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Natriumiodid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O