Merck
  • Effect of carob (Ceratonia siliqua L.) flour on the antioxidant potential, nutritional quality, and sensory characteristics of fortified durum wheat pasta.
Effect of carob (Ceratonia siliqua L.) flour on the antioxidant potential, nutritional quality, and sensory characteristics of fortified durum wheat pasta.
Food chemistry (2015-10-17)
Łukasz Sęczyk, Michał Świeca, Urszula Gawlik-Dziki
ZUSAMMENFASSUNG
This paper presents a study on the effect of carob flour addition from 1% to 5% (w/w) on phenolics content, antioxidant activity, nutritional quality, and sensory attributes of wheat pasta. An increase of about 2-folds, 18-folds and 3-folds in phenolics content, antiradical activity and reducing power for pasta fortified with 5% of carob flour was observed, respectively, compared to the control. Expected glycemic index (eGI) was increased proportionally to the substitution level and ranged between 72.2 and 83.9 for 1-5% of supplement, respectively. Furthermore, pasta fortification affected the in vitro bioaccessibility of nutrients. In case of 5% supplemented pasta, the digestibility of starch and protein decreased by about 9% compared to the control. The replacement of semolina with carob flour from 1% to 5% had no significant effect on pasta sensory attributes. In conclusion, carob flour seems to be a promising functional ingredient for pasta fortification.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Picrylsulfonsäure -Lösung, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
5-(Dimethylamino)-1-naphthalinsulfonamid, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.