Merck
  • A validated LC/UV method for the determination of four adulterating drugs in herbal slimming capsules.
A validated LC/UV method for the determination of four adulterating drugs in herbal slimming capsules.
Journal of pharmaceutical and biomedical analysis (2015-10-11)
Giacomo Russo, Francesco Barbato, Lucia Grumetto
ZUSAMMENFASSUNG
A simple LC/UV method for the simultaneous identification and quantification of Fluoxetine, Tiratricol, Benfluorex and Pseudoephedrine in slimming formulation is proposed. The method demonstrated effective in the analyses of herbal mixtures marketed in Italy as slimming capsules. Sixteen different herbal mixtures, selected among the most frequently compounded in Italian pharmacies, were tested as matrices. HPLC analyses were performed in a gradient mode on a C18 stationary phase The method was validated for accuracy, precision, linearity and selectivity. The limits of detection (LOD) were 3.4 μg/mL for Pseudoephedrine, 1.1 μg/mL for Triac, 0.9 μg/mL for Fluoxetine, and 0.8 μg/mL for Benfluorex. Repeatability and intermediate precision, expressed as percent of relative standard deviation, ranged from 3 to 7 and from 7 to 12, respectively. Given these limits, the developed method is proposed for the simple and cost effective screening of herbal products illegally adulterated with these four drugs known to enhance slimming effects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
Benzoesäure, ACS reagent, ≥99.5%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Sigma-Aldrich
Lidocain, powder
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
L-Ascorbinsäure, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)