Merck
Robust measurement of vitamin A status in plasma and blood dried on paper.
Prostaglandins, leukotrienes, and essential fatty acids (2015-10-23)
Yichao Huang, Peter Roy Clements, Robert Alan Gibson
ZUSAMMENFASSUNG
Vitamin A deficiency is the leading cause of preventable blindness in children and increases the risk of disease and death from severe infections. In addition, fat soluble vitamin A and associated retinoids directly regulate the expression of genes involved in fatty acid metabolism. Conventional methods for measuring vitamin A involve venipuncture, centrifugation and refrigeration all of which make measuring vitamin A in nutritional surveys expensive. We aimed to develop a simple and robust system for measurement of retinol (biomarker for vitamin A) using dried blood spot (DBS) samples. Low recoveries and inconsistent results reported by others were found to be due to poor extraction efficiency rather than retinol instability. Maintaining acid conditions during extraction resulted in recoveries >95% with <6.5% of coefficient of variation. Using isocratic high performance liquid chromatography, separation was achieved in <3.5 min. Detector response was linear (R(2)=0.9939) within a range of 0.05-2 μg/mL, with a limit of quantification of 0.05 μg/mL. Retinol in DBS was shown to be stable (>95%) at room temperature for up to 10 weeks. DBS values for retinol were highly correlated with venous blood samples from 24 healthy subjects (r=0.9724) and were consistent with results from a commercial laboratory. This simple and reliable method for the determination of vitamin A status should prove particularly valuable for population studies and large clinical trials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Butyliertes Hydroxytoluol, ≥99%, FCC, FG
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)