Merck
  • Monitoring Technology for Gamma-Aminobutyric acid Production in Polished Mochi Barley Grains using a Carbon Dioxide Sensor.
Monitoring Technology for Gamma-Aminobutyric acid Production in Polished Mochi Barley Grains using a Carbon Dioxide Sensor.
Journal of food science (2015-04-29)
Yasuo Watanabe, Kohki Kawata, Seiya Watanabe
ZUSAMMENFASSUNG
Gamma-aminobutyric acid (GABA) has many biological functions, including the inhibition of blood pressure increases and acceleration of growth hormone secretion. In this study, we discovered the utility of measuring the concentration of carbon dioxide (CO2 ) dissolved in the reaction solution, for development of a real-time and convenient technique to estimate GABA production. In addition to mochi barley bran, we examined the polished grains of three species: mochi barley (a variant of hulless barley), barley, and Japanese millet, all soaked in l-glutamic acid (l-Glu) solution at pH 4.5. We found a positive correlation between GABA and CO2 concentrations, and the production of CO2 was suppressed in the absence of l-Glu at pH 4.5. These results suggest that GABA content can be easily predicted by measuring the aqueous CO2 content using a CO2 sensor, during the process of GABA production in polished mochi barley grains and bran.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
L-Glutaminsäure, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
L-Glutaminsäure, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
Zitronensäure, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Zitronensäure, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Phthalsäure, ACS reagent, ≥99.5%
Sigma-Aldrich
L-Glutaminsäure, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Zitronensäure Trinatriumsalz, ≥98% (GC), anhydrous
Sigma-Aldrich
Zitronensäure, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
D-Glutaminsäure, ≥99% (TLC)
Sigma-Aldrich
2-Hydroxybuttersäure, 97%
Sigma-Aldrich
L-Glutaminsäure, FCC
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Phthalsäure, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Phthalsäure, reagent grade, 98%