Merck
Construct validation of the dietary inflammatory index among postmenopausal women.
Annals of epidemiology (2015-04-23)
Fred K Tabung, Susan E Steck, Jiajia Zhang, Yunsheng Ma, Angela D Liese, Ilir Agalliu, Melanie Hingle, Lifang Hou, Thomas G Hurley, Li Jiao, Lisa W Martin, Amy E Millen, Hannah L Park, Milagros C Rosal, James M Shikany, Nitin Shivappa, Judith K Ockene, James R Hebert
ZUSAMMENFASSUNG
Many dietary factors have either proinflammatory or anti-inflammatory properties. We previously developed a dietary inflammatory index (DII) to assess the inflammatory potential of diet. In this study, we conducted a construct validation of the DII based on data from a food frequency questionnaire and three inflammatory biomarkers in a subsample of 2567 postmenopausal women in the Women's Health Initiative Observational Study. We used multiple linear and logistic regression models, controlling for potential confounders, to test whether baseline DII predicted concentrations of interleukin-6, high-sensitivity C-reactive protein (hs-CRP), tumor necrosis factor alpha receptor 2, or an overall biomarker score combining all three inflammatory biomarkers. The DII was associated with the four biomarkers with beta estimates (95% confidence interval) comparing the highest with lowest DII quintiles as follows: interleukin-6: 1.26 (1.15-1.38), Ptrend < .0001; tumor necrosis factor alpha receptor 2: 81.43 (19.15-143.71), Ptrend = .004; dichotomized hs-CRP (odds ratio for higher vs. lower hs-CRP): 1.30 (0.97-1.67), Ptrend = .34; and the combined inflammatory biomarker score: 0.26 (0.12-0.40), Ptrend = .0001. The DII was significantly associated with inflammatory biomarkers. Construct validity of the DII indicates its utility for assessing the inflammatory potential of diet and for expanding its use to include associations with common chronic diseases in future studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Koffein, powder, ReagentPlus®
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Folsäure, ≥97%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Folsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Koffein, anhydrous, 99%, FCC, FG
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
Folsäure, meets USP testing specifications
Sigma-Aldrich
Koffein, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Koffein, meets USP testing specifications, anhydrous
Sigma-Aldrich
Koffein, BioXtra
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)