Merck
  • Microfluidic Mixing: A General Method for Encapsulating Macromolecules in Lipid Nanoparticle Systems.
Microfluidic Mixing: A General Method for Encapsulating Macromolecules in Lipid Nanoparticle Systems.
The journal of physical chemistry. B (2015-06-19)
Alex K K Leung, Yuen Yi C Tam, Sam Chen, Ismail M Hafez, Pieter R Cullis
ZUSAMMENFASSUNG
Previous work has shown that lipid nanoparticles (LNP) composed of an ionizable cationic lipid, a poly(ethylene glycol) (PEG) lipid, distearoylphosphatidylcholine (DSPC), cholesterol, and small interfering RNA (siRNA) can be efficiently manufactured employing microfluidic mixing techniques. Cryo-transmission electron microscopy (cryo-TEM) and molecular simulation studies indicate that these LNP systems exhibit a nanostructured core with periodic aqueous compartments containing siRNA. Here we examine first how the lipid composition influences the structural properties of LNP-siRNA systems produced by microfluidic mixing and, second, whether the microfluidic mixing technique can be extended to macromolecules larger than siRNA. It is shown that LNP-siRNA systems can exhibit progressively more bilayer structure as the proportion of bilayer DSPC lipid is increased, suggesting that the core of LNP-siRNA systems can exhibit a continuum of nanostructures depending on the proportions and structural preferences of component lipids. Second, it is shown that the microfluidic mixing technique can also be extended to encapsulation of much larger negatively charged polymers such mRNA (1.7 kb) or plasmid DNA (6 kb). Finally, as a demonstration of the generality of the microfluidic mixing encapsulation process, it is also demonstrated that negatively charged gold nanoparticles (5 nm diameter) can also be efficiently encapsulated in LNP containing cationic lipids. Interestingly, the nanostructure of these gold-containing LNP reveals a "currant bun" morphology as visualized by cryo-TEM. This structure is fully consistent with LNP-siRNA structure predicted by molecular modeling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Sigma-Aldrich
Natriumacetat, >99%, FG
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Natriumacetat, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumacetat -Lösung, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Zitronensäure Trinatriumsalz, ≥98% (GC), anhydrous
Sigma-Aldrich
Natriumacetat, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
MES -Lösung, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Ethansulfonsäure, 95%
Sigma-Aldrich
Natriumacetat, meets USP testing specifications, anhydrous
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Zitrat-konzentrierte Lösung -Lösung, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Natriumacetat, BioXtra, ≥99.0%
Sigma-Aldrich
L-α-Phosphatidylethanolamin, Dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications