Merck
  • Regioselective chemical and rapid enzymatic synthesis of a novel redox – Antiproliferative molecular hybrid.
Regioselective chemical and rapid enzymatic synthesis of a novel redox – Antiproliferative molecular hybrid.
European journal of medicinal chemistry (2015-04-16)
Elena Geromichalou, Nisar Sayyad, Eleni Kyriakou, Alexandra V Chatzikonstantinou, Efstathia Giannopoulou, Norbert Vrbjar, Haralabos P Kalofonos, Haralambos Stamatis, Andreas G Tzakos
ZUSAMMENFASSUNG
Recent science evidenced the interlinkage of oxidative stress and cancer. Due to the inherent complexity of cancer and its accompanying effect of oxidative stress, novel molecules, containing combinatorial functionalities should be targeted. Herein, we synthesized gemcitabine-5'-O-lipoate derived from a regioselective coupling of the chemotherapeutic drug gemcitabine (GEM), a first-line agent for cancer therapy and α-lipoic acid (LA), a potent antioxidant. gemcitabine-5'-O-lipoate was obtained in 4 chemical steps. To avoid the tedious and laborious chemical steps we also utilized biocatalysis using immobilized Candida antarctica lipase B (CALB), and the optimum conditions for the regioselective and one-pot synthesis of gemcitabine-5'-O-lipoate were established by exploiting different solvents (organic solvents and ionic liquids) and enzyme immobilization (acrylic resin and carbon nanotubes). Cytotoxic activity of co-administrating GEM and LA was proven to be synergistic against non-small cell lung cancer cells whereas antagonistic for bladder cancer cells. In contrast, the gemcitabine-5'-O-lipoate hybrid was found to be superior to the parent compounds against both non-small cell lung cancer and bladder cancer cells as also was found to preserve the redox activity of the parent compound LA. The regioselective biotransformation mediated synthesis of the anticancer-antioxidant hybrid illustrates the capacity of biocatalysis to act as an asset in molecular hybridization techniques.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
L-Cystein, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
4-(Dimethylamino)-pyridin, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Cystein, 97%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid, 99%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Di-tert-Butyldicarbonat, ReagentPlus®, 99%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
L-Cystein, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Natriumcarbonat, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Natriumcarbonat, BioXtra, ≥99.0%
Sigma-Aldrich
Diethylether
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
1-Butyl-3-methylimidazoliumtetrafluorborat, ≥98%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis