Merck
  • The deubiquitinating enzyme complex BRISC is required for proper mitotic spindle assembly in mammalian cells.
The deubiquitinating enzyme complex BRISC is required for proper mitotic spindle assembly in mammalian cells.
The Journal of cell biology (2015-07-22)
Kaowen Yan, Li Li, Xiaojian Wang, Ruisha Hong, Ying Zhang, Hua Yang, Ming Lin, Sha Zhang, Qihua He, Duo Zheng, Jun Tang, Yuxin Yin, Genze Shao
ZUSAMMENFASSUNG
Deubiquitinating enzymes (DUBs) negatively regulate protein ubiquitination and play an important role in diverse physiological processes, including mitotic division. The BRCC36 isopeptidase complex (BRISC) is a DUB that is specific for lysine 63-linked ubiquitin hydrolysis; however, its biological function remains largely undefined. Here, we identify a critical role for BRISC in the control of mitotic spindle assembly in cultured mammalian cells. BRISC is a microtubule (MT)-associated protein complex that predominantly localizes to the minus ends of K-fibers and spindle poles and directly binds to MTs; importantly, BRISC promotes the assembly of functional bipolar spindle by deubiquitinating the essential spindle assembly factor nuclear mitotic apparatus (NuMA). The deubiquitination of NuMA regulates its interaction with dynein and importin-β, which are required for its function in spindle assembly. Collectively, these results uncover BRISC as an important regulator of the mitotic spindle assembly and cell division, and have important implications for the development of anticancer drugs targeting BRISC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, Molecular Biology, ≥97.0%
Sigma-Aldrich
Thymidin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Thymidin, ≥99%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, ≥97.0%