Merck
Proteomic analysis of 3-MCPD and 3-MCPD dipalmitate toxicity in rat testis.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2015-06-15)
Stefanie Sawada, Axel Oberemm, Thorsten Buhrke, Christine Meckert, Christel Rozycki, Albert Braeuning, Alfonso Lampen
ZUSAMMENFASSUNG
Thermal treatment of foodstuff containing fats and salt promotes the formation of 3-chloropropane-1,2-diol (3-MCPD) and its fatty acid esters. 3-MCPD-exposed rats develop testicular lesions and Leydig cell tumors. 3-MCPD and 3-MCPD ester toxicity is thought to be caused by 3-MCPD and its metabolites, since 3-MCPD esters are hydrolyzed in the gut. Inhibition of glycolysis is one of the few known molecular mechanisms of 3-MCPD toxicity. To obtain deeper insight into this process, a comparative proteomic approach was chosen, based on a 28-days repeated-dose feeding study with male Wistar rats. Animals received equimolar doses of 3-MCPD or 3-MCPD dipalmitate. A lower dose of 3-MCPD dipalmitate was also administered. Absence of histopathological changes supported an analysis of early cellular disturbance. Testes were analyzed by two-dimensional gel electrophoresis followed by mass-spectrometric protein identification. Data provide a comprehensive overview of proteomic changes induced by 3-MCPD and 3-MCPD dipalmitate in rat testis in an early phase of organ impairment. Results are compatible with known 3-MCPD effects on reproductive function, substantially extend our knowledge about cellular responses to 3-MCPD and support the hypothesis that toxicity of 3-MCPD and 3-MCPD esters is mediated via common effectors. DJ-1 was identified as a candidate marker for 3-MCPD exposure.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Jodacetamid, BioUltra
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Jodacetamid, Single use vial of 56 mg
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Jodacetamid, ≥99% (NMR), crystalline
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Acrylamid, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Thioharnstoff, ACS reagent, ≥99.0%
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Spermin, ≥97%
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Acrylamid, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Thioharnstoff, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acrylamid -Lösung, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
(±)-3-Chlor-1,2-propandiol, 98%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Spermin, suitable for cell culture, BioReagent
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Spermin, ≥99.0% (GC)
Sigma-Aldrich
Harnstoff, meets USP testing specifications
SAFC
Jodacetamid
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O