Merck
  • Thin-layer chromatography/matrix-assisted laser desorption/ionisation mass spectrometry and matrix-assisted laser desorption/ionisation mass spectrometry imaging for the analysis of phospholipids in LS174T colorectal adenocarcinoma xenografts treated with the vascular disrupting agent DMXAA.
Thin-layer chromatography/matrix-assisted laser desorption/ionisation mass spectrometry and matrix-assisted laser desorption/ionisation mass spectrometry imaging for the analysis of phospholipids in LS174T colorectal adenocarcinoma xenografts treated with the vascular disrupting agent DMXAA.
Rapid communications in mass spectrometry : RCM (2015-09-26)
Afnan Batubara, Vikki A Carolan, Paul M Loadman, Chris Sutton, Steve D Shnyder, Malcolm R Clench
ZUSAMMENFASSUNG
5,6-Dimethylxanthenone-4-acetic acid (DMXAA) is a low molecular weight drug of the flavonoid group, which has an anti-vascular effect in tumours causing endothelial cell apoptosis and activation of cytokines. Flavonoid-based compounds have been reported to lead to an upregulation in the expression of lysophosphatidylcholines (LPC)-type lipids in solid tumours. A study employing TLC/MALDI-MS and MALDI-MS imaging to examine LS174T colorectal adenocarcinoma xenografts following administration of DMXAA has been conducted into this effect. LS174T colorectal adenocarcinoma xenografts grown in male immune-deficient mice were treated with 27.5 mg/kg DMXAA. The control (before treatment) and 4 h and 24 h post-treatment tumours were excised and divided into two. MALDI-MS imaging experiments were carried out on 12 µm cryosections sections taken from one half of the tumours and from the other half the lipids were extracted and analysed by TLC/MALDI-MS. These experiments were carried out in triplicate. Statistical analysis of the MALDI-MS imaging data set indicated an increased amount of LPC in the 24 h post-treated sample and a decreased amount of PC in the 24 h post-treated sample, compared with the 4 h post-treated sample and the control. These effects were confirmed by the TLC/MALDI-MS data. The lipid extracts were separated into six spots on the TLC plate. These were identified as arising from different lipids classes, i.e. LPC, sphingomyelins (SM), phosphatidylcholines (PC) and phosphatidylethanolamines (PE). The TLC/MALDI-MS data indicated that LPC were highly expressed in the 4 h and 24 h post-treated tumour samples compared with the control. Examination of the mass spectrometric images confirms this increase and demonstrates additionally that the increase in the signals arising from LPC appears to be localised primarily within the central areas of the xenograft. An increase in expression of LPC lipids in solid tumours treated with DMXAA has been demonstrated and shown to be localised in the central area of the tumour.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Lithiumchlorid -Lösung, 8 M, Molecular Biology, ≥99%
Sigma-Aldrich
Lithiumchlorid, Molecular Biology, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Lithiumchlorid, powder, ≥99.98% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Lithiumchlorid, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Sigma-Aldrich
Lithiumchlorid, BioUltra, Molecular Biology, anhydrous, ≥99.0% (AT)
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Lithiumchlorid, AnhydroBeads, −10 mesh, 99.998% trace metals basis