Merck
  • Role of iron oxide core of polymeric nanoparticles in the thermosensitivity of colon cancer cell line HT-29.
Role of iron oxide core of polymeric nanoparticles in the thermosensitivity of colon cancer cell line HT-29.
International journal of hyperthermia : the official journal of European Society for Hyperthermic Oncology, North American Hyperthermia Group (2015-05-12)
Elaheh Esmaelbeygi, Samideh Khoei, Sepideh Khoee, Samira Eynali
ZUSAMMENFASSUNG
In this study the effect of PLGA polymeric nanoparticles as a 5-fluorouracil (5-FU) carrier with and without iron oxide core and hyperthermia were investigated on the level of DNA damage in a spheroid culture model of HT-29 colon cancer cell lines by alkaline comet assay. First, HT-29 colon cancer cells were cultured in vitro as spheroids with a mean diameter of 100 µm. The spheroids were then treated with different concentrations of 5-FU or nanoparticles as 5-FU carriers with and without an iron oxide core for one volume-doubling time of the spheroids (71 h) and hyperthermia at 43 °C for 1 h. Finally, the effect of treatment on viability and level of DNA damage was examined using trypan blue dye exclusion assay and alkaline comet assay, respectively. Results showed that hyperthermia in combination with 5-FU or nanoparticles as 5-FU carriers significantly induced the most DNA damage as compared with the control group. The extent of DNA damage following treatment with 5-FU-loaded nanoparticles combined with hyperthermia was significantly more than for 5-FU combined with hyperthermia. In comparison to the effect of 5-FU-loaded nanoparticles with the iron oxide core and 5-FU-loaded nanoparticle without the iron oxide core, the nanoparticles with the iron oxide core combined with hyperthermia induced more DNA damage than the nanoparticles without the iron oxide core. According to this study, hyperthermia is a harmful agent and nanoparticles are effective delivery vehicles for drugs into colon cancer cells. The iron oxide filled nanoparticles increased the effect of the hyperthermia. All these factors have a significant role in the treatment of colorectal cancer cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Phenylessigsäure, 99%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Phenylessigsäure, ≥99%, FCC, FG
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis