Merck
  • Simple surface engineering of polydimethylsiloxane with polydopamine for stabilized mesenchymal stem cell adhesion and multipotency.
Simple surface engineering of polydimethylsiloxane with polydopamine for stabilized mesenchymal stem cell adhesion and multipotency.
Scientific reports (2015-12-10)
Yon Jin Chuah, Yi Ting Koh, Kaiyang Lim, Nishanth V Menon, Yingnan Wu, Yuejun Kang
ZUSAMMENFASSUNG
Polydimethylsiloxane (PDMS) has been extensively exploited to study stem cell physiology in the field of mechanobiology and microfluidic chips due to their transparency, low cost and ease of fabrication. However, its intrinsic high hydrophobicity renders a surface incompatible for prolonged cell adhesion and proliferation. Plasma-treated or protein-coated PDMS shows some improvement but these strategies are often short-lived with either cell aggregates formation or cell sheet dissociation. Recently, chemical functionalization of PDMS surfaces has proved to be able to stabilize long-term culture but the chemicals and procedures involved are not user- and eco-friendly. Herein, we aim to tailor greener and biocompatible PDMS surfaces by developing a one-step bio-inspired polydopamine coating strategy to stabilize long-term bone marrow stromal cell culture on PDMS substrates. Characterization of the polydopamine-coated PDMS surfaces has revealed changes in surface wettability and presence of hydroxyl and secondary amines as compared to uncoated surfaces. These changes in PDMS surface profile contribute to the stability in BMSCs adhesion, proliferation and multipotency. This simple methodology can significantly enhance the biocompatibility of PDMS-based microfluidic devices for long-term cell analysis or mechanobiological studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Fluoreszeindiacetat, used as cell viability stain
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Propidiumjodid -Lösung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.