Merck
  • Quality by design approach for formulation, evaluation and statistical optimization of diclofenac-loaded ethosomes via transdermal route.
Quality by design approach for formulation, evaluation and statistical optimization of diclofenac-loaded ethosomes via transdermal route.
Pharmaceutical development and technology (2014-02-05)
Shashank Jain, Niketkumar Patel, Parshotam Madan, Senshang Lin
ZUSAMMENFASSUNG
The objective of this study was to fabricate and understand ethosomal formulations of diclofenac (DF) for enhanced anti-inflammatory activity using quality by design approach. DF-loaded ethosomal formulations were prepared using 4 × 5 full-factorial design with phosphatidylcholine:cholesterol (PC:CH) ratios ranging between 50:50 and 90:10, and ethanol concentration ranging between 0% and 30% as formulation variables. These formulations were characterized in terms of physicochemical properties and skin permeation kinetics. The interaction of formulation variables had a significant effect on both physicochemical properties and permeation kinetics. The results of multivariate regression analysis illustrated that vesicle size and elasticity of ethosomes were the dominating physicochemical properties affecting skin permeation, and could be suitably controlled by manipulation of formulation variables to optimize the formulation and enhance the skin permeation of DF-loaded ethosomes. The optimized formulation had ethanol concentration of 22.9% and PC:CH ratio of 88.4:11.6, with vesicle size of 144 ± 5 nm, zeta potential of -23.0 ± 3.76 mV, elasticity of 2.48 ± 0.75 and entrapment efficiency of 71 ± 4%. Permeation flux for the optimized formulation was 12.9 ± 1.0 µg/h cm(2), which was significantly higher than the drug-loaded conventional liposome, ethanolic or aqueous solution. The in vivo study indicated that optimized ethosomal hydrogel exhibited enhanced anti-inflammatory activity compared with liposomal and plain drug hydrogel formulations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Di-(N-succinimidyl)-carbonat, ≥95%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Rhodamin 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Rhodamin 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Krypton-84Kr, 90 atom %
Sigma-Aldrich
Krypton-84Kr, 80 atom %