Merck
  • Analytical method for the determination and a survey of parabens and their derivatives in pharmaceuticals.
Analytical method for the determination and a survey of parabens and their derivatives in pharmaceuticals.
Environmental research (2015-08-09)
Cristina Moreta, María-Teresa Tena, Kurunthachalam Kannan
ZUSAMMENFASSUNG
Exposure of humans to parabens is a concern due to the estrogenic activity of these compounds. Parabens are widely used as preservatives in some personal care products, foodstuffs and pharmaceuticals owing to their low cost, high water solubility and broad spectrum antimicrobial properties. Despite this, little is known on the occurrence of parabens in pharmaceutical products. In this study, a method based on solid-liquid or liquid-liquid extraction (SLE or LLE), and high performance liquid chromatography (HPLC) coupled with triple quadrupole tandem mass spectrometry (QqQ or MS/MS) was developed for the determination of six most frequently used parabens and four paraben derivatives (methyl- and ethyl-protocatechuates, and mono- and di-hydroxybenzoic acids) in pharmaceuticals. A sample-purification step involving solid phase extraction (SPE) was optimized for the analysis of solid and lipid-rich pharmaceuticals. To our knowledge, this is the first comprehensive report on the occurrence of parabens in pharmaceuticals. The developed method was applied for the analysis of 128 liquid/syrup, cream, solid, prescription or over-the counter (OTC) drugs collected from the USA and a few other countries in Europe and Asia. Although majority of the drugs analyzed in the study did not contain parabens, concentrations as high as 2 mg/g were found in some drugs. Methyl- and propyl- parabens were the frequently detected compounds. 4-Hydroxybenzoic acid was the major metabolite found in pharmaceutical products.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
4-Hydroxybenzoesäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
4-Hydroxybenzoesäure, ReagentPlus®, 99%
Sigma-Aldrich
3,4-Dihydroxy-benzoesäure, ≥97.0% (T)
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Protocatechusäureethylester, 97%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.