Merck
  • Micro-solid-phase extraction of organochlorine pesticides using porous metal-organic framework MIL-101 as sorbent.
Micro-solid-phase extraction of organochlorine pesticides using porous metal-organic framework MIL-101 as sorbent.
Journal of chromatography. A (2015-05-20)
Zhenzhen Huang, Hian Kee Lee
ZUSAMMENFASSUNG
In this study, a metal-organic framework material, MIL-101, used as a micro-solid-phase extraction (μ-SPE) sorbent for efficient enrichment of five organochlorine pesticides (OCPs), including α-HCH, Aldrin, α-Chlordane, Dieldrin and p,p'-DDD from water samples, followed by gas chromatography-mass spectrometry, is reported. This study demonstrated a new application of MIL-101 using μ-SPE, an advantage of the latter being its ability to process complex aqueous matrices, due to the protection of the sorbent afforded by the hollow fiber membrane bag. Key factors affecting extraction efficiency were studied, including elution solvent, extraction and desorption time. Under the optimal extraction conditions, the calibration plots were linear from 0.05 to 50 ng/mL for α-HCH and p,p'-DDD, and 0.1 to 50 ng/mL for the other three analytes. The limits of detection were between 0.0025 and 0.016 ng/mL. The relative recoveries of the OCPs spiked into real water samples (at 5 ng/mL of each analyte) ranged from 87.6 to 98.6% with relative standard deviations of <10%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Terephthalsäure, 98%
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Sigma-Aldrich
Natriumacetat, >99%, FG
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Natriumacetat, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumacetat -Lösung, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Natriumacetat, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Natriumacetat, meets USP testing specifications, anhydrous
Sigma-Aldrich
Ethylacetat
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Natriumacetat, BioXtra, ≥99.0%
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.