Merck
  • Polo‑like kinase 3 inhibits osteosarcoma cell proliferation and tumorigenesis via cooperative interaction with p21.
Polo‑like kinase 3 inhibits osteosarcoma cell proliferation and tumorigenesis via cooperative interaction with p21.
Molecular medicine reports (2015-09-10)
Honglin Lv, Guangting Gao, Linlin Zhang, Yujie Sun
ZUSAMMENFASSUNG
Polo‑like kinase 3 (Plk3) is a member of the Plk family. It is dysregulated in certain types of cancer, including colorectal and pancreatic cancer. However, the expression status and biological function of Plk3 in osteosarcoma (OS) remain poorly understood. Following evaluation of the role of Plk3 in OS, the present study indicates that Plk3 is downregulated in OS cell lines and tissues, and increased expression levels of Plk3 are associated with improved rates of overall survival of patients. In addition, to investigate the role of Plk3 in cell proliferation and tumorigenicity in vitro, two recombinant lentiviruses expressing Plk3 short hairpin RNA, as well as a recombinant plasmid carrying Plk3, were developed and transfected into Saos‑2 and U2OS cells, respectively. Cell cycle analysis by flow cytometry demonstrated the influence of Plk3 on the arrest of cell cycle progression at the G1 phase. Following knock down of Plk3, the growth and colony formation of Saos‑2 cells increased significantly, whereas the overexpression of Plk3 resulted in the opposite trend. Furthermore, a 5‑ethynyl‑2'‑deoxyuridine assay, using U2OS cell lines, indicated the same tendency. The in vivo interaction between Plk3 and p21 in Saos‑2 cells was detected and the protein level of p21 was observed to be consistent with that of Plk3. These results imply that Plk3 is involved in the inhibition of cell proliferation and tumorigenesis, which may occur via interactions with p21, thus, Plk3 may be considered as a potential candidate for targeted therapy of OS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Deoxycholsäure Natriumsalz, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Deoxycholsäure Natriumsalz, ≥97% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Propidiumjodid -Lösung