Merck
  • Pharmacokinetic evaluation of the equivalency of gavage, dietary, and drinking water exposure to manganese in F344 rats.
Pharmacokinetic evaluation of the equivalency of gavage, dietary, and drinking water exposure to manganese in F344 rats.
Toxicological sciences : an official journal of the Society of Toxicology (2015-03-01)
Melanie L Foster, Thomas B Bartnikas, Laura C Johnson, Carolina Herrera, Michael A Pettiglio, Athena M Keene, Michael D Taylor, David C Dorman
ZUSAMMENFASSUNG
Concerns exist as to whether individuals may be at greater risk for neurotoxicity following increased manganese (Mn) oral intake. The goals of this study were to determine the equivalence of 3 methods of oral exposure and the rate (mg Mn/kg/day) of exposure. Adult male rats were allocated to control diet (10 ppm), high manganese diet (200 ppm), manganese-supplemented drinking water, and manganese gavage treatment groups. Animals in the drinking water and gavage groups were given the 10 ppm manganese diet and supplemented with manganese chloride (MnCl(2)) in drinking water or once-daily gavage to provide a daily manganese intake equivalent to that seen in the high-manganese diet group. No statistically significant difference in body weight gain or terminal body weights was seen. Rats were anesthetized following 7 and 61 exposure days, and samples of bile and blood were collected. Rats were then euthanized and striatum, olfactory bulb, frontal cortex, cerebellum, liver, spleen, and femur samples were collected for chemical analysis. Hematocrit was unaffected by manganese exposure. Liver and bile manganese concentrations were elevated in all treatment groups on day 61 (relative to controls). Increased cerebellum manganese concentrations were seen in animals from the high-manganese diet group (day 61, relative to controls). Increased (relative to all treatment groups) femur, striatum, cerebellum, frontal cortex, and olfactory bulb manganese concentrations were also seen following gavage suggesting that dose rate is an important factor in the pharmacokinetics of oral manganese. These data will be used to refine physiologically based pharmacokinetic models, extending their utility for manganese risk assessment by including multiple dietary exposures.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Mangan, powder, ≥99.9% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Mangan, powder, −325 mesh, ≥99% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Mangan, chips, thickness <2.0 mm, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salpeter-14N-Säurelösung, ~10 N in H2O, 99.99 atom % 14N
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, rod, 10mm, diameter 4.0mm, cast, 99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, rod, 50mm, diameter 4.0mm, cast, 99.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Mangan, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 98.7%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.