Merck
Ghrelin receptor modulates T helper cells during intestinal inflammation.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2015-08-01)
M Di Giovangiulio, N Stakenborg, G Bosmans, E Meroni, G Farro, P J Gomez-Pinilla, I Depoortere, G E Boeckxstaens, G Matteoli
ZUSAMMENFASSUNG
The orexigenic peptide ghrelin has anti-inflammatory properties in colitis, however, the mechanism of action and the immune cells targeted remain still to be elucidated. Here, we assessed the possible effect of ghrelin on T helper (Th) cells in a T cell transfer model of chronic colitis. Disease was induced in the recombination activating gene 1 knockout mice (Rag1(-/-) ) by adoptive transfer of naïve Th cells from ghrelin receptor knockout mice (GRLN-R(-/-) ) or littermate wild-type (WT) mice. The course and severity of colitis was assessed by monitoring body weight, diarrhea score, histological analysis, gene expression, and flow cytometry analysis. The possible effects of ghrelin on Th cell proliferation, polarization, and apoptosis was examined in vitro. Our data showed that Rag1(-/-) mice injected with GRLN-R(-/-) Th cells displayed increased severity of colitis compared to mice injected with WT Th cells. In addition, Rag1(-/-) mice injected with GRLN-R(-/-) Th cells had significantly higher intestinal inflammation and increased accumulation of Th1 and Th17 cells in the colon. In vitro, ghrelin directly affected proliferation of Th cells and induced apoptosis whereas it did not influence Th cell polarization. Our observations suggest that ghrelin modulates Th effector cells in the gut controlling proliferation and inducing apoptosis. Our findings further support the use of ghrelin as a novel therapeutic option to treat intestinal inflammatory diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
SAFC
L-Glutamin
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Interleukin-6 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-6 from mouse, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture, carrier free
Sigma-Aldrich
IL-6 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Interleukin-6 from rat, IL-6, recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture