Merck
Analyte-induced photoreduction method for visual and colorimetric detection of tyrosine.
Analytica chimica acta (2015-05-24)
Elumalai Satheeshkumar, Jyisy Yang
ZUSAMMENFASSUNG
A new method based on photochemical formation of silver nanoparticles (AgNPs) was developed for detection of tyrosine (Tyr). To selectively detect Tyr and to simplify the detection procedure, the photoactivity of Tyr was utilized to trigger the photochemical reduction in production of AgNPs. The drastic change of solution color caused by the surface plasmon resonance (SPR) absorption band of the formed AgNPs was used to extract the quantitative information of Tyr. This developed method is simple in detection, while both the sensitivity and selectivity are significant improved. Meanwhile, the solution color was changed from colorless to dark yellow after the formation of AgNPs, which allows a much higher sensitivity in visual identification when compared with the SPR band shifting technique commonly, used in conventional colorimetric methods. To optimize the detection system and to understand the mechanism in this proposed method, parameters such as irradiation time, intensity of light source, and the concentration of Tyr were systematically examined. Results indicated that these factors mainly affected the reaction rate of photoreduction. The morphologies of the formed AgNPs were similar, but with small differences in particle sizes. In the examination of selectivity, sixteen other amino acids were examined. Results indicated that only amino acids of tryptophan, cysteine and histidine are photoactive and possess potential interferences in analysis of Tyr. Quantitative studies indicated that a linear response up to 10 μM with a detection limit of 100 nM could be obtained. For visually detection, color change could be observed with a concentration as low as 500 nM of Tyr.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
SAFC
Glycin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kaliumhydroxid, ≥99.95% trace metals basis
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Methionin, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Phenylalanin, ReagentPlus®, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, 99%, FCC
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Prolin, ReagentPlus®, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Alanin, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Methionin, 99%, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Cystein, technical grade
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Valin, ReagentPlus®, ≥99.0% (NT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Alanin, ≥99%, FCC, FG
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
N-Phenylglycin, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Histidin, ≥99% (TLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Methionin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
DL-Valin, ≥97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Salpeter-14N-Säurelösung, ~10 N in H2O, 99.99 atom % 14N
Preise und Verfügbarkeit sind derzeit nicht verfügbar.