Merck
Oxidative DNA damage induced by di-(2-ethylhexyl) phthalate in HEK-293 cell line.
Environmental toxicology and pharmacology (2015-04-23)
Xuan Wang, Lijie Jiang, Lan Ge, Min Chen, Guang Yang, Fang Ji, Laifu Zhong, Yingjie Guan, Xiaofang Liu
ZUSAMMENFASSUNG
Di-(2-ethylhexyl) phthalate (DEHP) is commonly employed as a plasticizer. We have found that exposure of human embryonic kidney cell line 293 (HEK-293) to DEHP resulted in a crucial dose-dependent increase of DNA strand breaks in a comet assay. To elucidate the role of glutathione (GSH) in the DNA damage, the cells were pretreated with buthionine-(S,R)-sulfoximine (BSO) and pretreated with N-acetylcysteine (NAC), a GSH precursor. Here we show that depletion of GSH in HEK-293 cells with BSO dramatically increased the susceptibility of HEK-293 cells to DEHP-induced DNA damage. Furthermore, when the intracellular GSH content was elevated by NAC, the DNA damage induced by DEHP was almost completely abolished. In addition, DEHP had effect on lysosomal or mitochondrial damage at high dose level. These results indicate that DEHP exerts genotoxic effects in HEK-293 cells, probably through DNA damage induced by oxidative stress; GSH is responsible for cellular defense against DEHP-induced DNA damage; lysosome and mitochondria may be the vital targets in DEHP-induced DNA damage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
N-Acetyl-L-Cystein, BioReagent, suitable for cell culture
Sigma-Aldrich
1-Methyl-2-pyrrolidon, anhydrous, 99.5%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
N-Acetyl-L-Cystein, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Phthaldialdehyd, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Phthaldialdehyd, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Ethidiumbromid, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Phthaldialdehyd, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
Rhodamin 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
N-Acetyl-L-Cystein, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Rhodamin 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Ethidiumbromid, ~95% (HPLC)
Sigma-Aldrich
Acridinorange -hydrochlorid -Lösung, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Ethidiumbromid -Lösung, suitable for fluorescence, ~1% in H2O
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)