- The anti-DNA antibody: origin and impact, dogmas and controversies.
The anti-DNA antibody: origin and impact, dogmas and controversies.
Nature reviews. Rheumatology (2015-06-03)
Ole P Rekvig
PMID26034836
ZUSAMMENFASSUNG
The inclusion of 'the anti-DNA antibody' by the ACR and the Systemic Lupus International Collaborating Clinics (SLICC) as a criterion for systemic lupus erythematosus does not convey the diverse origins of these antibodies, whether their production is transient or persistent (which is heavily influenced by the nature of the inducing antigens), the specificities exerted by these antibodies or their clinical impact-or lack thereof. A substantial amount of data not considered in clinical medicine could be added from basic immunology evidence, which could change the paradigms linked to what 'the anti-DNA antibody' is, in a pathogenic, classification or diagnostic context.