Merck
  • Ultra-high performance liquid chromatography-electrospray tandem mass spectrometry for the analysis of antibiotic residues in environmental waters.
Ultra-high performance liquid chromatography-electrospray tandem mass spectrometry for the analysis of antibiotic residues in environmental waters.
Environmental science and pollution research international (2015-06-25)
Qiang Xue, Yanjie Qi, Fei Liu
ZUSAMMENFASSUNG
An optimized solid-phase extraction (SPE) and ultra-high performance liquid chromatography-electrospray tandem mass spectrometry (UPLC-MS/MS) method was developed for the effective analysis of 35 antibiotics including sulfonamides (SAs), quinolones (QLs), tetracyclines (TCs), macrolides (MALs), lincomycin (LIN), and chloramphenicol (CAP). The addition of 0.1% formic acid to the mobile phase was favorable for the formation of [M + H](+) and the enhancement in the detection signals, but using ammonium formate decreased [M + H](+) with a corresponding reduction in the response of CAP. The optimal pH range for the SPE was 4.5 ∼ 5.0 with 6 mL aqueous ammonia/methanol (5/95, v/v) as the optimized eluent. An internal standard (IS) was selected for each type of analytes based on similarities in classification and retention time. The detection was completed in less than 10 min and was excellent with method detection limits (MDL) of 0.29 ∼ 4.03 ng/L. The recoveries of the antibiotics in samples from ultrapure water and groundwater were 67.13 ∼ 93.00% and 68.91 ∼ 92.67%, respectively. The antibiotics in samples collected from wastewater, surface water, and groundwater were also effectively detected. This newly developed method has the advantages of short detection times, small sample consumption, excellent reproducibility, and high sensitivity. This provides a reliable and promising technique for the simultaneous detection and monitoring of various residual antibiotics in aqueous environmental samples.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Ammoniumformat, reagent grade, 97%
Sigma-Aldrich
Doxorubicin -hydrochlorid, 98.0-102.0% (HPLC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ammoniumformat, ≥99.995% trace metals basis
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammoniumformiat -Lösung, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin -hydrochlorid, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Nalidixinsäure, ≥98%
Sigma-Aldrich
Decylamin, 95%
Sigma-Aldrich
2,3-Diaminonaphthalin, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)
Sigma-Aldrich
2,3-Diaminonaphthalin, ≥95% (HPLC), powder
Sigma-Aldrich
Erythromycin-Standard -Lösung, 1 mg/mL in H2O
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Oxolinsäure, quinolone antibiotic
Sigma-Aldrich
Decylamin, ≥99.0% (GC)
Sigma-Aldrich
Josamycin, ≥90% (HPLC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)