Merck
  • Combined PCSK9 and APOE polymorphisms are genetic risk factors associated with elevated plasma lipid levels in a Thai population.
Combined PCSK9 and APOE polymorphisms are genetic risk factors associated with elevated plasma lipid levels in a Thai population.
Lipids (2015-04-23)
Nutjaree Jeenduang, Sureerut Porntadavity, Smith Wanmasae
ZUSAMMENFASSUNG
Proprotein convertase subtilisin/kexin type 9 (PCSK9) and apolipoprotein E (ApoE) play a key role in the regulation of lipid metabolism. We aimed to investigate the effects of PCSK9 (R46L, I474V, and E670G) and APOE polymorphisms on lipid levels in a Southern Thai population. A total of 495 participants (307 urban, 188 rural) were recruited for the study. Anthropometric and biochemical variables were evaluated. PCSK9 and APOE polymorphisms were analyzed using PCR-RFLP. The 46L urban male carriers had significantly higher diastolic blood pressure (DBP) and fasting blood sugar compared with non-carriers. In contrast, the 46L urban female carriers had significantly lower total cholesterol (TC) and LDL-C levels compared with non-carriers. The 474V rural female carriers had significantly lower HDL-C levels than non-carriers. The 670G urban female carriers showed significantly higher TC and LDL-C levels compared with non-carriers. APOE4 carriers had increased TC and LDL-C levels relative to APOE3 carriers in the urban males. APOE2 carriers had decreased TC and/or LDL-C levels compared with APOE3 carriers in urban males and females. A significant trend of increased TC and LDL-C levels was observed in non-APOE4-PCSK9 670EE carriers to APOE4-PCSK9 670EG carriers in urban subjects. In summary, R46L, I474V, and E670G may be genetic risk factors for cardiovascular disease (CVD) in urban males, rural females, and urban females, respectively. In contrast, R46L had a favorable lipid profiles that may protect against CVD in urban females. The combination of PCSK9 E670G and APOE polymorphisms may represent an independent factor for the determination of lipid levels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®