Merck
  • Advances in Use of Capsule-Based Fluorescent Sensors for Measuring Acidification of Endocytic Compartments in Cells with Altered Expression of V-ATPase Subunit V1G1.
Advances in Use of Capsule-Based Fluorescent Sensors for Measuring Acidification of Endocytic Compartments in Cells with Altered Expression of V-ATPase Subunit V1G1.
ACS applied materials & interfaces (2015-06-19)
Maria De Luca, Marzia M Ferraro, Raimo Hartmann, Pilar Rivera-Gil, Andreas Klingl, Moritz Nazarenus, Agnese Ramirez, Wolfgang J Parak, Cecilia Bucci, Rosaria Rinaldi, Loretta L del Mercato
ZUSAMMENFASSUNG
Acidification of eukaryotic cell compartments is accomplished by vacuolar H+-ATPases (V-ATPases), large multisubunit complexes able to pump protons into the lumen of organelles or in the extracellular medium. V-ATPases are involved in a number of physiological cellular processes, and thus regulation of V-ATPase activity is of crucial importance for the cell. Indeed, dysfunction of V-ATPase or alterations of acidification have been recently recognized as key factors in a variety of human diseases. In this study, we applied capsule-based pH sensors and a real-time tracking method for investigating the role of the V1G1 subunit of V-ATPases in regulating the activity of the proton pump. We first constructed stable cell lines overexpressing or silencing the subunit V1G1. Second, we used fluorescent capsule-based pH sensors to monitor acidification before and during internalization by modified and control living cells. By using a simple real-time method for tracking capsule internalization, we were able to identify different capsule acidification levels with respect to each analyzed cell and to establish the kinetics for each. The intracellular pH measurements indicate a delay in acidification in either V1G1-overexpressing or V1G1-silenced cells compared to controls. Finally, in an independent set of experiments, we applied transmission electron microscopy and confocal fluorescence microscopy to further investigate the internalization of the capsules. Both analyses confirm that capsules are engulfed in acidic vesicular structures in modified and control cell lines. The use of capsule-based pH sensors allowed demonstration of the importance of the V1G1 subunit in V-ATPase activity concerning intravesicular acidification. We believe that the combined use of these pH-sensor system and such a real-time method for tracking their internalization path would contribute to systematically measure the proton concentration changes inside the endocytic compartments in various cell systems. This approach would provide fundamental information regarding molecular mechanisms and factors that regulate intracellular acidification, vesicular trafficking, and cytoskeletal reorganizations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamin -Lösung, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Dulbecco Modifiziertes Eagle-Medium – hoher Glucosegehalt, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Natriumcarbonat, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Natriumcarbonat, BioXtra, ≥99.0%
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Phenolrot, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Phenolrot, ACS reagent
Sigma-Aldrich
Rhodamin-B-isothiocyanat, mixed isomers
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Rhodamin-B-Isothiocyanat, BioReagent, mixed isomers, suitable for protein labeling
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Ethansulfonsäure, 95%
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)