Merck
  • Rapid high performance liquid chromatography-high resolution mass spectrometry methodology for multiple prenol lipids analysis in zebrafish embryos.
Rapid high performance liquid chromatography-high resolution mass spectrometry methodology for multiple prenol lipids analysis in zebrafish embryos.
Journal of chromatography. A (2015-08-19)
Chiara Martano, Vera Mugoni, Federica Dal Bello, Massimo M Santoro, Claudio Medana
ZUSAMMENFASSUNG
The analysis of lipid molecules in living organism is an important step in deciphering metabolic pathways. Recently, the zebrafish has been adopted as a valuable animal model system to perform in vivo metabolomics studies, however limited methodologies and protocols are currently available to investigate zebrafish lipidome and even fewer to analyze specific classes of lipids. Here we present an HPLC-HRMS based method to rapidly measure multiple prenol lipid molecules from zebrafish tissues. In particular, we have optimized our method for concurrent detection of ubiquinones (Coenzyme Q6, Coenzyme Q9, Coenzyme Q10), cholesterol, vitamin E (α-tocopherol), vitamin K1 and vitamin K2. The purpose of this study was to compare different ionization modes, mobile phases and stationary phases in order to optimize lipid molecules separation. After HPLC-HRMS parameters selection, several extraction conditions from zebrafish embryos were evaluated. We assessed our methodology by quantitation of analytical recovery on zebrafish extracts from wild-type or zebrafish mutants (barolo) affected by impaired biosynthesis of ubiquinones.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
α-Tocopherol, ≥95.5%
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Coenzym Q10, ≥98% (HPLC)
Sigma-Aldrich
Heptan, anhydrous, 99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Vitamin K1, viscous liquid
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Sigma-Aldrich
Natriumacetat, >99%, FG
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixture of isomers
Sigma-Aldrich
Lithiumacetat, 99.95% trace metals basis
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Sigma-Aldrich
Natriumacetat, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Natriumacetat -Lösung, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Natriumacetat, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
3-Methyl-2-buten-1-ol, 99%
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
Natriumacetat, meets USP testing specifications, anhydrous
Sigma-Aldrich
Coenzym Q9, ≥96.0% (HPLC)