Merck
  • Differences in gene expression and alterations in cell cycle of acute myeloid leukemia cell lines after treatment with JAK inhibitors.
Differences in gene expression and alterations in cell cycle of acute myeloid leukemia cell lines after treatment with JAK inhibitors.
European journal of pharmacology (2015-08-25)
Pawel Gunerka, Barbara Dymek, Aleksandra Stanczak, Anna Bujak, Paulina Grygielewicz, Pawel Turowski, Karolina Dzwonek, Monika Lamparska-Przybysz, Tadeusz Pietrucha, Maciej Wieczorek
ZUSAMMENFASSUNG
Janus kinase (JAK) inhibitors are a promising treatment strategy in several hematological malignancies and autoimmune diseases. A number of inhibitors are in clinical development, and two have already reached the market. Unfortunately, all of them are burdened with different toxicity profiles. To check if the JAK inhibitors of different selectivity evoke different responses on JAK2-dependent and independent cells, we have used three acute myeloid leukemia cell lines with confirmed JAK2 mutation status. We have found that JAK inhibitors exert distinct effect on the expression of BCLXL, CCND1 and c-MYC genes, regulated by JAK pathway, in JAK2 wild type cells in comparison to JAK2 V617F-positive cell lines. Moreover, cell cycle analysis showed that inhibitors alter the cycle by arresting cells in different phases. Our results suggest that observed effect of JAK2 inhibitors on transcription and cell cycle level in different cell lines are associated not with activity within JAK family, but presumably with other off-target activities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®