Merck
  • A Delicate Balance When Substituting a Small Hydrophobe onto Low Molecular Weight Polyethylenimine to Improve Its Nucleic Acid Delivery Efficiency.
A Delicate Balance When Substituting a Small Hydrophobe onto Low Molecular Weight Polyethylenimine to Improve Its Nucleic Acid Delivery Efficiency.
ACS applied materials & interfaces (2015-10-24)
Deniz Meneksedag-Erol, Remant Bahadur KC, Tian Tang, Hasan Uludağ
ZUSAMMENFASSUNG
High molecular weight (HMW) polyethylenimine (PEI) is one of the most versatile nonviral gene vectors that was extensively investigated over the past two decades. The cytotoxic profile of HMW PEI, however, encouraged a search for safer alternatives. Because of lack of cytotoxicity of low molecular weight (LMW) PEI, enhancing its performance via hydrophobic modifications has been pursued to this end. Since the performance of modified PEIs depends on the nature and extent of substituents, we systematically investigated the effect of hydrophobic modification of LMW (1.2 kDa) PEI with a short propionic acid (PrA). Moderate enhancements in PEI hydrophobicity resulted in enhanced cellular uptake of polyplexes and siRNA-induced silencing efficacy, whereas further increase in PrA substitution abolished the uptake as well as the silencing. We performed all-atom molecular dynamics simulations to elucidate the mechanistic details behind these observations. A new assembly mechanism was observed by the presence of hydrophobic PrA moieties, where PrA migrated to core of the polyplex. This phenomenon caused higher surface hydrophobicity and surface charge density at low substitutions, and it caused deleterious effects on surface hydrophobicity and cationic charge at higher substitutions. It is evident that an optimal balance of hydrophobicity/hydrophilicity is needed to achieve the desired polyplex properties for an efficient siRNA delivery, and our mechanistic findings should provide valuable insights for the design of improved substituents on nonviral carriers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Diethylether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Diethylether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
N-Hydroxysuccinimid, 98%
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Diethylether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Diethylether
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Diethylether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Diethylether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in acetic acid
Sigma-Aldrich
5-Carboxyfluorescein, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%