Merck
  • Determination of Antiproliferative Activities of Volatile Contents and HPLC Profiles of Dicranum scoparium (Dicranaceae, Bryophyta).
Determination of Antiproliferative Activities of Volatile Contents and HPLC Profiles of Dicranum scoparium (Dicranaceae, Bryophyta).
Combinatorial chemistry & high throughput screening (2015-03-06)
Gökhan Abay, Muhammed Altun, Serkan Koldaş, Ali Rıza Tüfekçi, Ibrahim Demirtas
ZUSAMMENFASSUNG
The aim of this study was to examine the anticancer activities and phytochemical profiles of Dicranum scoparium against HeLa cell lines. The bio-guided fractionation studies of dichloromethane extract have high antiproliferative activities. Fractions 7, 9, 19, 20 are rich source of unsaturated fatty acids, and- in the case of Fr-19 may improve the antiproliferative activities as well as increase the unsaturated fatty acid content. The effect of proliferative activities in hexane extract can be attributed to the saturated fatty acid composition of D. scoparium. The Fr-9 exhibited strong antiproliferative activity at concentrations of 100 and 50 μg mL(-1) compared to 5-FU. The fractions of 7, 9, 19 and 20 from dichloromethane extracts exhibited antiproliferative activities at a concentration of 100 μg mL(-1). The HPLC-TOF/MS studies gave nine compounds from the most active fraction of dichloromethane at concentrations of 250 and 100 μg mL(-1). The lower activities were obtained from the fractions including steroid derivatives.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Kaliumhydroxid, ≥99.95% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Cer, ingot, under oil, 99.9% trace rare earth metals basis
Sigma-Aldrich
Cer, chips
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®
Cerium, rod, 50mm, diameter 6.35mm, cast, 99.9%
Cerium, rod, 100mm, diameter 12.7mm, cast, 99.9%
Cerium, rod, 50mm, diameter 12.7mm, cast, 99.9%
Cerium, rod, 100mm, diameter 6.35mm, cast, 99.9%
Cerium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.9%