Merck
  • Functional Reconstruction of Tracheal Defects by Protein-Loaded, Cell-Seeded, Fibrous Constructs in Rabbits.
Functional Reconstruction of Tracheal Defects by Protein-Loaded, Cell-Seeded, Fibrous Constructs in Rabbits.
Tissue engineering. Part A (2015-06-23)
Lindsey M Ott, Cindy H Vu, Ashley L Farris, Katrina D Fox, Richard A Galbraith, Mark L Weiss, Robert A Weatherly, Michael S Detamore
ZUSAMMENFASSUNG
Tracheal stenosis is a life-threatening disease and current treatments include surgical reconstruction with autologous rib cartilage and the highly complex slide tracheoplasty surgical technique. We propose using a sustainable implant, composed of a tunable, fibrous scaffold with encapsulated chondrogenic growth factor (transforming growth factor-beta3 [TGF-β3]) or seeded allogeneic rabbit bone marrow mesenchymal stromal cells (BMSCs). In vivo functionality of these constructs was determined by implanting them in induced tracheal defects in rabbits for 6 or 12 weeks. The scaffolds maintained functional airways in a majority of the cases, with the BMSC-seeded group having an improved survival rate and the Scaffold-only group having a higher occurrence of more patent airways as determined by microcomputed tomography. The BMSC group had a greater accumulation of inflammatory cells over the graft, while also exhibiting normal epithelium, subepithelium, and cartilage formation. Overall, it was concluded that a simple, acellular scaffold is a viable option for tracheal tissue engineering, with the intraoperative addition of cells being an optional variation to the scaffolds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
Sigma-Aldrich
1,1,1,3,3,3-Hexafluor-2-propanol, ≥99%
Sigma-Aldrich
Eisen(III)-chlorid, reagent grade, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Pikrinsäure, moistened with water, ≥98%
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Milchsäure, meets USP testing specifications
Sigma-Aldrich
Phosphatgepufferte Kochsalzlösung -Lösung, BioPerformance Certified, pH 7.4
Sigma-Aldrich
Glycolsäure, ReagentPlus®, 99%
Sigma-Aldrich
Eisen(III)-chlorid, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Zitronensäure, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Milchsäure, 88%, FCC
Sigma-Aldrich
Pikrinsäure -Lösung, 1.3% in H2O (saturated)
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Milchsäure, natural, ≥85%
Sigma-Aldrich
Milchsäure -Lösung, ACS reagent, ≥85%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Natriumthiosulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Eisen(III)-chlorid, ≥99.9% trace metals basis
Sigma-Aldrich
Säurefuchsin, used in tissue staining
Sigma-Aldrich
Eosin Y -Lösung, 5 wt. % in H2O
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Zitronensäure, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Sigma-Aldrich
Eosin Y Dinatriumsalz, certified by the BSC
Sigma-Aldrich
Eosin Y Dinatriumsalz, ≥85 % by HPLC