Merck
  • Analysis of the metabolic profile of parishin by ultra-performance liquid chromatography/quadrupole-time of flight mass spectrometry.
Analysis of the metabolic profile of parishin by ultra-performance liquid chromatography/quadrupole-time of flight mass spectrometry.
Biomedical chromatography : BMC (2015-05-27)
Chunlan Tang, Li Wang, Jiejiang Li, Xinxin Liu, Mengchun Cheng, Hongbin Xiao
ZUSAMMENFASSUNG
Parishin is a dominant active ingredient originating from Gastrodia elata Blume, and has good neuroprotective effects against brain disorders. In the present study, the metabolic profile of parishin by in vitro and in vivo experiments was investigated using ultra-high performance liquid chromatography coupled with quadrupole-time of flight mass spectrometry (UHPLC/Q-TOF MS) combined with an automated MS(E) technique. By comparison with reference compounds, accurate mass measurement, the characteristic fragmentation patterns of the parent drug parishin and gastrodin and relevant bio-transformation knowledge, 14 metabolites (seven hydrolyzates and seven derivatives of gastrodin) were detected and identified in rat plasma and urine after intragastric administration of parishin, including processes of hydrolyzation, oxidation, sulfation and glucuronidation. According to the proposed metabolic pathways of parishin, in vitro hydrolytic experiments and metabolic study of gastrodin in rat plasma, it can be inferred that parishin mainly functions as a prodrug and undergoes hydrolysis before being absorbed into the blood. The hydrolyzate, mainly gastrodin, was involved in further metabolism, which was responsible for pharmacological activities of parishin. In conclusion, this work provides valuable information on parishin metabolism using a rapid and reliable UHPLC/Q-TOF MS method, which could be widely used for the metabolic investigation of natural product.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitril, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitril, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitril, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.