Merck
  • Synergistic antitumor effects of S-1 with eribulin in vitro and in vivo for triple-negative breast cancer cell lines.
Synergistic antitumor effects of S-1 with eribulin in vitro and in vivo for triple-negative breast cancer cell lines.
SpringerPlus (2014-08-21)
Masato Terashima, Kazuko Sakai, Yosuke Togashi, Hidetoshi Hayashi, Marco A De Velasco, Junji Tsurutani, Kazuto Nishio
ZUSAMMENFASSUNG
Triple-negative breast cancer (TNBC) is associated with a higher incidence of recurrence and distant metastasis and a poor prognosis, whereas effective treatment strategies remain to be established. Finding an effective treatment for TNBC has become imperative. We examined the effect of the combination of S-1 (or 5-FU in an in vitro study) and eribulin in TNBC cell lines. The in vitro effect of the combination was examined in four TNBC cell lines (MDA-MB-231, MDA-MB-468, BT-549 and MX-1) using a combination index and isobolograms. In addition, we assessed the effect of the combination in an MDA-MB-231 tumor xenograft model. A synergistic effect was observed in three TNBC cell lines (MDA-MB-231, MDA-MB-468, and MX-1), and in an in vivo study, the combination of S-1 and eribulin resulted in significantly higher antitumor effects compared with S-1 or eribulin alone. 5-FU induced epithelial-mesenchymal transition (EMT) change in the TNCB cell line, as supported by the decreased expression of epithelial marker and the increased expression of mesenchymal markers. Meanwhile, TGF-beta induced EMT changes in a TNBC cell line and decreased the sensitivity to 5-FU. This result suggests that 5-FU-induced EMT changes reduce the sensitivity to 5-FU. In contrast, eribulin induced a mesenchymal-epithelial transition (MET) in a TNBC cell line. The EMT phenotype induced by 5-FU was also canceled by eribulin. We demonstrate that the combination of S-1 (5-FU) and eribulin exerts a synergistic effect for TNBC cell lines through MET-induction by eribulin. Therefore, this combination therapy may be a potential treatment option for TNBC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Natriumorthovanadat, ≥90% (titration)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumorthovanadat, 99.98% trace metals basis
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Kalium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Kaliumhydrid, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Natriumfluorid, 99.99% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumfluorid, BioXtra, ≥99%
Sigma-Aldrich
Natriumfluoridlösung
Sigma-Aldrich
Natriumfluorid, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Kaliumhydrid, in paraffin
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl