Merck
TAOK3 phosphorylates the methylenecyclopropane nucleoside MBX 2168 to its monophosphate.
Antiviral research (2015-04-11)
Gloria Komazin-Meredith, Steven C Cardinale, Katelyn Comeau, Kevin J Magalhaes, Caroll B Hartline, John D Williams, Timothy J Opperman, Mark N Prichard, Terry L Bowlin
ZUSAMMENFASSUNG
Monohydroxymethyl methylenecyclopropane nucleosides (MCPNs) with ether or thioether substituents at the 6-position show promise as broad-spectrum herpes virus inhibitors. Their proposed mechanism of action involves sequential phosphorylation to a triphosphate, which can then inhibit viral DNA polymerase. The inhibition of herpes simplex virus (HSV) by these compounds is not dependent on the viral thymidine kinase (TK), which is known to phosphorylate acyclovir (ACV), a standard treatment for HSV infections. Previous studies on the mechanism of action of these compounds against human cytomegalovirus (HCMV) implicated a host kinase in addition to HCMV UL97 kinase in performing the initial phosphorylation. After first eliminating other candidate HSV-1 encoded kinases (UL13 and US3) as well as potential host nucleoside kinases, using activity-based fractionation, we have now identified the host serine-threonine protein kinase TAOK3 as the kinase responsible for transforming the representative monohydroxymethyl MCPN analog MBX 2168 to its monophosphate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
L-Threonin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Phosphoenolbrenztraubensäure Trinatriumsalz Hydrat, ≥97% (enzymatic)
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
L-Threonin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
SAFC
L-Threonin
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ganciclovir, ≥99% (HPLC), powder
Sigma-Aldrich
L-Threonin, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
DL-Serin, ≥98% (TLC)
Sigma-Aldrich
Acycloguanosin, ≥99% (HPLC), powder
Sigma-Aldrich
DL-Serin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)