Merck
  • An improved method for retrospective quantification of sulfur mustard exposure by detection of its albumin adduct using ultra-high pressure liquid chromatography-tandem mass spectrometry.
An improved method for retrospective quantification of sulfur mustard exposure by detection of its albumin adduct using ultra-high pressure liquid chromatography-tandem mass spectrometry.
Analytical and bioanalytical chemistry (2015-07-15)
ChangCai Liu, LongHui Liang, Yu Xiang, HuiLan Yu, ShiKun Zhou, HaiLing Xi, ShiLei Liu, JingQuan Liu
ZUSAMMENFASSUNG
Sulfur mustard (HD) adduct to human serum albumin (ALB) at Cys-34 residue has become an important and long-term retrospective biomarker of HD exposure. Here, a novel, sensitive, and convenient approach for retrospective quantification of HD concentration exposed to plasma was established by detection of the HD-ALB adduct using ultra-high pressure liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) with a novel non-isotope internal standard (IS). The HD-ALB adduct was isolated from HD-exposed plasma with blue Sepharose. The adduct was digested with proteinase K to form sulfur-hydroxyethylthioethyl ([S-HETE])-Cys-Pro-Phe tripeptide biomarker. The tripeptide adduct could be directly analyzed by UHPLC-MS/MS without an additional solid phase extraction (SPE), which was considered as a critical procedure in previous methods. The easily available 2-chloroethyl ethylsulfide (2-CEES) as HD surrogate was first reported to be used as IS in place of traditional d8-HD for quantification of HD exposure. Furthermore, 2-CEES was also confirmed to be a good IS alternative for quantification of HD exposure by investigation of product ion spectra for their corresponding tripeptide adducts which exhibited identical MS/MS fragmentation behaviors. The method was found to be linear between 1.00 and 250 ng•mL(-1) HD exposure (R(2)>0.9989) with precision of <4.50% relative standard deviation (%RSD), accuracy range between 96.5% and 114%, and a calculated limit of detection (LOD) of 0.532 ng•mL(-1). The lowest reportable limit (LRL) is 1.00 ng•mL(-1), over seven times lower than that of the previous method. The entire method required only 0.1 mL of plasma sample and took under 7 h without special sample preparation equipment. It is proven to be a sensitive, simple, and rugged method, which is easily applied in international laboratories to improve the capabilities for the analysis of biomedical samples related to verification of the Chemical Weapon Convention (CWC).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Natriumphosphat, 96%
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Bromkresolgrün, ACS reagent, Dye content 95 %
Sigma-Aldrich
Schwefel, powder, 99.98% trace metals basis
Sigma-Aldrich
Schwefel, 99.998% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Trizma® Base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® Base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Trizma® Base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
2-Chlorethylethylsulfid, 97%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture