Merck
  • Improved Natural Killer cell activity and retained anti-tumor CD8(+) T cell responses contribute to the induction of a pathological complete response in HER2-positive breast cancer patients undergoing neoadjuvant chemotherapy.
Improved Natural Killer cell activity and retained anti-tumor CD8(+) T cell responses contribute to the induction of a pathological complete response in HER2-positive breast cancer patients undergoing neoadjuvant chemotherapy.
Journal of translational medicine (2015-06-28)
E Muraro, E Comaro, R Talamini, E Turchet, G Miolo, S Scalone, L Militello, D Lombardi, S Spazzapan, T Perin, S Massarut, D Crivellari, Riccardo Dolcetti, D Martorelli
ZUSAMMENFASSUNG
Locally advanced HER2-overexpressing breast cancer (BC) patients achieve a high rate of pathological complete responses (pCR) after neoadjuvant chemotherapy (NC). The apparently unaltered immune proficiency of these patients together with the immune-modulating activities of NC drugs suggest a potential contribution of host immunity in mediating clinical responses. We thus performed an extensive immunomonitoring in locally advanced BC patients undergoing NC to identify immunological correlates of pCR induction. The immune profile of 40 HER2-positive and 38 HER2-negative BC patients was characterized at diagnosis and throughout NC (Paclitaxel and Trastuzumab, or Docetaxel and Epirubicin, respectively). The percentages of circulating immune cell subsets including T and B lymphocytes, Natural Killer (NK) cells, regulatory T cells, T helper 17 lymphocytes, were quantified by multiparametric flow cytometry. NK cells functional activity was evaluated through the analysis of NF-kB nuclear translocation by Multispectral flow cytometry, and with the in vitro monitoring of Trastuzumab-mediated antibody-dependent cell cytotoxicity (ADCC). CD8(+) T cell responses against six different tumor-associated antigens (TAA) were characterized by IFN-γ ELISPOT and IFN-γ/IL-2 DualSpot assays. After NC, HER2-positive patients showed a significant increase in the number of NK cells and regulatory T cells irrespective of the pathological response, whereas patients undergoing a pCR disclosed higher percentages of T helper 17 cells. Notably, a significant increase in the number of activated NK cells was observed only in HER2-positive patients achieving a pCR. Characterization of anti-tumor T cell responses highlighted sustained levels of CD8(+) T cells specific for survivin and mammaglobin-A throughout NC in patients undergoing a pCR in both arms. Moreover, HER2-positive patients achieving a pCR were characterized by a multi-epitopic and polyfunctional anti-tumor T cell response, markedly reduced in case of partial response. These results indicate that maintenance of functional T cell responses against selected antigens and improvement of NK cell proficiency during NC are probably critical requirements for pCR induction, especially in HER2-positive BC patients. Trail registration: NCT02307227, registered on ClinicalTrials.gov ( http://www.clinicaltrials.gov , November 26, 2014).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
SAFC
L-Glutamin
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Calcein-AM -Lösung, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®