Merck
  • Enniatin-containing solutions for oromucosal use: Quality-by-design ex-vivo transmucosal risk assessment of composition variability.
Enniatin-containing solutions for oromucosal use: Quality-by-design ex-vivo transmucosal risk assessment of composition variability.
International journal of pharmaceutics (2015-06-28)
Lien Taevernier, Sven Detroyer, Lieselotte Veryser, Bart De Spiegeleer
ZUSAMMENFASSUNG
Fusafungine, a mixture of the cyclic hexadepsipeptides enniatins, is currently on the market for the treatment of upper respiratory tract diseases because of its bacteriostatic and anti-inflammatory effects. In this study, a quality-by-design risk assessment was performed with two objectives: (i) investigate whether enniatins are able to permeate the mucosa and reach blood circulation, as the summary of product characteristics indicates this is not the case, and if so, to quantify their transmucosal kinetics and (ii) study the influence of excipient concentration variability on mucosal permeation. First, the concentration of the two main excipients isopropyl myristate and ethanol, known penetration enhancers, in several marketed samples was determined using GC-FID. Then, the transmucosal kinetics of the enniatins were quantitatively evaluated for different dose solutions, using porcine buccal mucosa in an ex-vivo in-vitro Franz diffusion cell set-up, with UHPLC-MS/MS bioanalytics. This study demonstrated that enniatins are capable of permeating the mucosa. However, no risk of a significant different transmucosal permeability with varying excipient concentrations was detected.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
N,N-Dimethylacetamid, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylacetamid, anhydrous, 99.8%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
N,N-Dimethylacetamid, suitable for peptide synthesis, ≥99.8% (GC)
SAFC
N,N-Dimethylacetamid, Ph. Eur.
Sigma-Aldrich
N,N-Dimethylacetamid-d9, 99 atom % D
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®